Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900004

Sigma-Aldrich

Propyl 4-hydroxybenzoate

Vetec, reagent grade, 98%

Synonym(s):

4-Hydroxybenzoic acid propyl ester, Propyl paraben

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2CH2CH2CH3
CAS Number:
Molecular Weight:
180.20
Beilstein/REAXYS Number:
1103245
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

95-98 °C (lit.)

SMILES string

CCCOC(=O)c1ccc(O)cc1

InChI

1S/C10H12O3/c1-2-7-13-10(12)8-3-5-9(11)6-4-8/h3-6,11H,2,7H2,1H3

Inchi Key

QELSKZZBTMNZEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

356.0 °F

flash_point_c

180 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis G Maki et al.
Critical care medicine, 39(4), 613-620 (2011-01-05)
Catheter-related bloodstream infection is the greatest threat to the safety of patients on hemodialysis. Catheter lock solutions containing heparin have been linked to an increased risk of hemorrhage and thrombocytopenia. To ascertain the safety and efficacy for prevention of catheter-related
C L Barberis et al.
Journal of food protection, 73(8), 1493-1501 (2010-09-08)
Each year, a significant portion of the peanuts produced cannot be marketed because of fungal disease at the postharvest stage and mycotoxin contamination. Antioxidants could be used as an alternative to fungicides to control ochratoxigenic fungi in peanuts during storage.
Miguel González-Doncel et al.
Environmental pollution (Barking, Essex : 1987), 184, 360-369 (2013-10-08)
Here we proposed a battery of non-invasive biomarkers and a histological survey to examine physiological/anatomical features in embryos, eleutheroembryos (13 days post-fertilization, dpf), and larvae (28-42 dpf) of medaka to investigate the effects of embryonic exposure to propylparaben (PrP). Concentrations <1000 μg
Aeshna Amin et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 75(2), 206-212 (2010-03-09)
p-Hydroxybenzoic acid esters (parabens) are commonly used antimicrobial preservatives in pharmaceutical formulations. Two microorganisms, isolated from non-sterile methyl paraben (MP) and propyl paraben (PP) solutions, were found to degrade the respective parabens. Identification by 16S rRNA partial gene sequencing revealed
O Herrero et al.
Mutation research, 743(1-2), 20-24 (2012-01-18)
Di(2-ethylhexyl)phthalate, triclosan and propylparaben are contaminants of emerging concern that have been subjected to extensive toxicological studies, but for which limited information is currently available concerning adverse effects on terrestrial plant systems. The Allium cepa test, which is considered one

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service