Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900003

Sigma-Aldrich

Benzotriazole

Vetec, reagent grade, 98%

Synonym(s):

1,2,3-Benzotriazole, 1H-Benzotriazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N3
CAS Number:
Molecular Weight:
119.12
Beilstein/REAXYS Number:
112133
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

4.1 (vs air)

vapor pressure

0.04 mmHg ( 20 °C)

product line

Vetec

assay

98%

mp

97-99 °C (lit.)

SMILES string

c1ccc2[nH]nnc2c1

InChI

1S/C6H5N3/c1-2-4-6-5(3-1)7-9-8-6/h1-4H,(H,7,8,9)

Inchi Key

QRUDEWIWKLJBPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

338.0 °F - closed cup

flash_point_c

170 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rozanna Avagyan et al.
Environmental science and pollution research international, 22(8), 5842-5849 (2014-10-25)
Textiles play an important role in our daily life, and textile production is one of the oldest industries. In the manufacturing chain from natural and/or synthetic fibers to the final clothing products, the use of many different chemicals is ubiquitous.
K Pavić et al.
European journal of medicinal chemistry, 86, 502-514 (2014-09-10)
Novel primaquine semicarbazides 7a-l and ureas 9a-g with modified benzhydryl, trityl, phenyl or hydroxyalkyl substituents were prepared and evaluated for cytostatic and antioxidative activities. Two synthetic approaches for preparation of the title semicarbazides were applied, both having certain advantages. In
Paul Thellmann et al.
Environmental science and pollution research international, 22(21), 16405-16416 (2014-11-14)
In order to evaluate surface water and the sediment quality of rivers connected to wastewater treatment plants (WWTPs) with different treatment technologies, fish embryo tests (FET) with Danio rerio were conducted using native water and sediment samples collected upstream and
Evangelia Terzopoulou et al.
Environmental science and pollution research international, 22(2), 1095-1112 (2014-08-12)
A multi-residue method, based on gas chromatography coupled to tandem mass spectrometry (GC-MS/MS), has been developed for the determination of 70 organic micropollutants from various chemical classes (organochlorinated, organophosphorous, triazines, carbamate and urea, polycyclic aromatic hydrocarbons, polychlorinated biphenyls, pharmaceuticals, phenols
Chiranjit Dutta et al.
ACS applied materials & interfaces, 7(33), 18397-18405 (2015-08-05)
Designing an effective peptide based molecular transporter for the intracellular delivery of hydrophilic therapeutic biomacromolecules remains a considerable challenge. Highly basic oligoarginine and lipidated arginine rich cell penetrating peptides have been reported in the literature as molecular transporters, which were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service