Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900002

Sigma-Aldrich

Dimethylglyoxime

Vetec, reagent grade

Synonym(s):

2,3-Butanedione dioxime, Diacetyldioxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(=NOH)C(=NOH)CH3
CAS Number:
Molecular Weight:
116.12
Beilstein/REAXYS Number:
506731
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder

mp

240-241 °C (lit.)

SMILES string

CC(=N/O)\C(C)=N\O

InChI

1S/C4H8N2O2/c1-3(5-7)4(2)6-8/h7-8H,1-2H3/b5-3+,6-4+

InChI key

JGUQDUKBUKFFRO-GGWOSOGESA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Sol. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Capeness et al.
New biotechnology, 32(6), 727-731 (2015-02-18)
Desulfovibrio alaskensis G20 is an anaerobic sulfate reducing bacteria. While Desulfovibrio species have previously been shown to reduce palladium and platinum to the zero-state, forming nanoparticles in the process; there have been no reports that D. alaskensis is able to
Jean-Marie Lachapelle et al.
Dermatology (Basel, Switzerland), 209(4), 288-290 (2004-11-13)
To determine the release of nickel from 1- and 2-euro coins and the ability to produce allergic contact dermatitis from the application of coins to the palmar skin of nickel-sensitized individuals. Three experiments were conducted. Experiments 1 and 2 checked
Carsten R Hamann et al.
Contact dermatitis, 62(4), 232-240 (2010-03-20)
China and Thailand produce large amounts of jewellery that are sold domestically and abroad. To identify nickel release and metal content in earrings purchased in China and Thailand. A total of 557 earrings were randomly purchased from vendors in 11
Jacob Pontoppidan Thyssen et al.
Journal of the American Academy of Dermatology, 58(6), 1000-1005 (2008-04-15)
Nickel sensitization is frequent among US patients with dermatitis and in the general population. In Europe, decreasing prevalences of nickel sensitization are observed as a result of the European Union Nickel Directive. However, no directive exists in the United States.
Katherine E Heim et al.
Contact dermatitis, 60(2), 100-105 (2009-02-12)
Cutaneous nickel allergy in the very young is not well documented or characterized. A significant number of individuals are nickel sensitized by their mid-teenage years. Recent studies suggest that children may become sensitized to nickel at an early age. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service