Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900001

Sigma-Aldrich

Eugenol

Vetec, reagent grade, 98%

Synonym(s):

2-Methoxy-4-(2-propenyl)phenol, 4-Allyl-2-methoxyphenol, 4-Allylguaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(H2C=CHCH2)C6H3-2-(OCH3)OH
CAS Number:
Molecular Weight:
164.20
Beilstein/REAXYS Number:
1366759
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.541 (lit.)

bp

254 °C (lit.)

mp

−12-−10 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

SMILES string

COc1cc(CC=C)ccc1O

InChI

1S/C10H12O2/c1-3-4-8-5-6-9(11)10(7-8)12-2/h3,5-7,11H,1,4H2,2H3

InChI key

RRAFCDWBNXTKKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

233.6 °F - closed cup

flash_point_c

112 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhavini Shah et al.
International journal of food microbiology, 161(1), 53-59 (2012-12-25)
There has been great interest in intervention strategies based on plant essential oils to control pathogens such as Escherichia coli O157:H7 and Listeria monocytogenes (Lm). However, the poor solubility of essential oils in water makes it difficult to disperse evenly
Katharina Bücher et al.
Clinical oral investigations, 19(7), 1653-1662 (2014-12-31)
This retrospective study analyzed restoration survival of composite fillings in children with at high caries risk in relation to age, sex, operator, tooth type, filling extension, and material used. Among 667 children treated in 2004-2012 in a university setting without
Hyang Nam et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 55, 106-112 (2013-01-15)
The oxidative damage of lipid, protein and DNA is known to be involved in chronic inflammation as well as metastasis. It has been highlighted for searching natural compounds without toxicity to prevent development of these diseases. Thus, it was investigated
I U Fischer et al.
Journal of chromatography, 525(2), 369-377 (1990-02-23)
The high-performance liquid chromatographic assay described permitted a simple, rapid, sensitive, selective and precise quantitative determination of eugenol in body fluids (serum, urine and bile) without derivatization. Amounts in the range 0.02-100 micrograms of eugenol per millilitre of body fluid
Seiichiro Fujisawa et al.
Toxicology, 177(1), 39-54 (2002-07-20)
To clarify the possible link between radicals and the cytotoxicity of eugenol-related compounds, 2-allyl-4-X-phenols (2-allyl-4-chlorophenol (1), 2-allyl-4-phenylphenol (2), 2-allyl-4-methoxyphenol (3), 2-allyl-4-acetylphenol (4), 2-allyl-4-nitrophenol (5), 2-allyl-4-t-butylphenol (6), 2-allyl-4-methyphenol (7), 2-allyl-4-bromophenol (8), 2,4-dimethoxyphenol (9)), and dimeric compounds from eugenol (4-allyl-2-methoxyphenol), BHA (2-t-butyl-4-methoxyphenol)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service