Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800452

Sigma-Aldrich

Zinc sulfate heptahydrate

LR, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99%
grade:
LR
Pricing and availability is not currently available.

grade

LR

product line

Vetec

assay

≥99%

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dervis Özcelik et al.
Biological trace element research, 150(1-3), 342-349 (2012-10-12)
Zinc is an element that under physiological conditions preferentially binds to and is a potent inducer of metallothionein under physiological conditions. The present study was conducted to explore whether zinc supplementation morphologically and biochemically protects against diabetic nephropathy through modulation
O I Smian et al.
Likars'ka sprava, (3-4)(3-4), 67-71 (2013-01-30)
The article analyzed 60 patients with community-aquired pneumonia from ages 6 to 18 years. The dependences of trace levels of zinc and copper in the serum of these children on the activity of the inflammatory process. Analysis of survey results
Zhixiang Xu et al.
Neuroscience letters, 541, 269-274 (2013-03-05)
Constitutive adult neurogenesis mainly occurs in the subgranular zone (SGZ) and subventricular zone (SVZ) of the mammalian brain. Intrinsic and extrinsic factors regulate adult neurogenesis processes including cell proliferation, migration, integration and neural survival. The potential contribution of olfactory sensory
Priyanga Ranasinghe et al.
Trials, 14, 52-52 (2013-02-21)
The number of people with diabetes is increasing worldwide, especially in developing South Asian countries. Therefore, preventing diabetes at the early stages has become an important issue. Recent clinical trials, systematic reviews, and meta-analysis have shown that zinc has beneficial
Na Yuan et al.
Bioscience, biotechnology, and biochemistry, 76(11), 2014-2020 (2012-11-08)
As the second most abundant transition metal in humans, zinc plays essential roles in normal cellular biological functions, including metabolism, signalling, proliferation, gene expression and apoptosis. We use ZnSO(4) as a stressor in this study to investigate for the first

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service