Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800451

Sigma-Aldrich

Zinc sulfate heptahydrate

AR, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99.5%
grade:
AR
form:
crystalline powder
Pricing and availability is not currently available.

grade

AR

product line

Vetec

assay

≥99.5%

form

crystalline powder

pH

4.4-5.6 (5% in water)

anion traces

chloride (Cl-): ≤0.0005%

cation traces

Fe: ≤0.0005%
heavy metals (as Pb): ≤0.001%

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priyanga Ranasinghe et al.
Trials, 14, 52-52 (2013-02-21)
The number of people with diabetes is increasing worldwide, especially in developing South Asian countries. Therefore, preventing diabetes at the early stages has become an important issue. Recent clinical trials, systematic reviews, and meta-analysis have shown that zinc has beneficial
O I Smian et al.
Likars'ka sprava, (3-4)(3-4), 67-71 (2013-01-30)
The article analyzed 60 patients with community-aquired pneumonia from ages 6 to 18 years. The dependences of trace levels of zinc and copper in the serum of these children on the activity of the inflammatory process. Analysis of survey results
Haichen Niu et al.
Pharmacology, biochemistry, and behavior, 103(3), 526-534 (2012-10-30)
Previous studies have shown that olfactory impairment by disrupting the olfactory epithelium prior to morphine administration attenuated the development addiction-related behaviors. However, it is unclear whether olfactory impairment will affect the expression of already established addiction-related behaviors. To address this
Laurent Dortet et al.
Antimicrobial agents and chemotherapy, 56(12), 6437-6440 (2012-10-17)
A biochemical test (Carba NP test II) was developed to identify carbapenemase production in Enterobacteriaceae and Pseudomonas spp. and to discriminate between the different types of carbapenemases (classes A, B, and D). It is based on the detection of the
Akash Malik et al.
Pediatrics, 132(1), e46-e52 (2013-06-05)
Zinc supplementation during diarrhea substantially reduces the incidence and severity of diarrhea. However, the effect of short-course zinc prophylaxis has been observed only in children >12 months of age. Because the incidence of diarrhea is comparatively high in children aged

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service