Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V800448

Sigma-Aldrich

Zinc

LR, dust, ≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
assay:
≥90%
grade:
LR
form:
dust
Pricing and availability is not currently available.

grade

LR

vapor pressure

1 mmHg ( 487 °C)

product line

Vetec

assay

≥90%

form

dust

resistivity

5.8 μΩ-cm, 20°C

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Casper Groth et al.
Development (Cambridge, England), 140(14), 3018-3027 (2013-06-21)
Developmental patterning requires the precise interplay of numerous intercellular signaling pathways to ensure that cells are properly specified during tissue formation and organogenesis. The spatiotemporal function of the Notch signaling pathway is strongly influenced by the biosynthesis and intracellular trafficking
A viewpoint about the treatment of Wilson's disease.
Abdul Qayyum Rana et al.
The Canadian journal of neurological sciences. Le journal canadien des sciences neurologiques, 40(4), 612-614 (2013-06-22)
I A Popov et al.
Molekuliarnaia biologiia, 47(3), 498-504 (2013-07-31)
Alzheimer's disease, is a lethal neurodegenerative pathology, characterized by the formation of soluble neurotoxic oligomers of the human amyloid-beta peptide Abeta which get accumulated forming polymeric extracellular aggregates (so-called amyloid plaques). The isomerized at aspartate 7 isoform of the human
Marco Ranocchiari et al.
Chimia, 67(6), 397-402 (2013-08-16)
Structural features, synthesis, and reactivity of Zn-biphenyl metal-organic frameworks with MOF-5 topology are presented to show the chemical flexibility of such materials and to demonstrate the challenges that can be encountered and solved to avoid interpenetration. We introduce the synthesis
Cigdem Karakukcu et al.
Clinical laboratory, 59(5-6), 557-562 (2013-07-20)
The aim of this study was to assess the acute and chronic effects of vigorous physical activity, with and without zinc supplementation, on distribution of elements in young amateur boxers. The 8 week intervention trial experiment was designed with 32

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service