Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800445

Sigma-Aldrich

Xylenes

LR, sulfur free, ≥98%

Synonym(s):

Xylene mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
LR
assay:
≥98%
bp:
137-140 °C (lit.)
vapor pressure:
18 mmHg ( 37.7 °C)
Pricing and availability is not currently available.

grade

LR

vapor pressure

18 mmHg ( 37.7 °C)

product line

Vetec

assay

≥98%

form

liquid

expl. lim.

7 %

dilution

(for analytical testing)

refractive index

n20/D 1.497 (lit.)

bp

137-140 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)cc1.Cc2cccc(C)c2.Cc3ccccc3C

InChI

1S/3C8H10/c1-7-3-5-8(2)6-4-7;1-7-4-3-5-8(2)6-7;1-7-5-3-4-6-8(7)2/h3*3-6H,1-2H3

InChI key

MVZVDAGWAAZJPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

77.0 °F - closed cup

flash_point_c

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-France le Rolle et al.
Journal of translational medicine, 13, 199-199 (2015-06-25)
Clinical therapy for metastatic colorectal cancer (CRC) remains limited, especially when the tumor harbors a KRAS mutation. This study aimed to identify prognostic biomarkers in CRC that are accessible for therapeutic inhibition. Conditioned media from human CRC epithelial cells and
Ryan P Smith et al.
Human gene therapy, 26(6), 377-385 (2015-03-27)
Much research has gone into the development of hybrid gene delivery systems that combine the broad tropism and efficient transduction of adenoviral vectors with the ability to achieve stable expression of cargo genes. In addition to gene therapy applications, such
Osceola Whitney et al.
Developmental neurobiology, 75(7), 778-790 (2014-11-20)
The forkhead domain FOXP2 and FOXP1 transcription factors are implicated in several cognitive disorders with language deficits, notably autism, and thus play a central role in learned vocal motor behavior in humans. Although a similar role for FoxP2 and FoxP1
Qiming Wang et al.
Nature communications, 5, 4899-4899 (2014-09-17)
Cephalopods can display dazzling patterns of colours by selectively contracting muscles to reversibly activate chromatophores--pigment-containing cells under their skins. Inspired by this novel colouring strategy found in nature, we design an electro-mechano-chemically responsive elastomer system that can exhibit a wide
Lian He et al.
Journal of environmental sciences (China), 23(1), 160-165 (2011-04-12)
The activities of CeO2 nanocubes calcined at different temperatures were tested for catalytic oxidation of o-xylene. Using CeO2 nanocubes as catalysts, complete catalytic oxidation of o-xylene was achieved below 210 degrees C. The CeO2 nanomaterials were characterized by means of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service