Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800438

Sigma-Aldrich

Triethanolamine

LR, ≥99%

Synonym(s):

2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)3N
CAS Number:
Molecular Weight:
149.19
Beilstein/REAXYS Number:
1699263
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

LR

vapor density

5.14 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

product line

Vetec

assay

≥99%

autoignition temp.

600 °F

expl. lim.

8.5 %

refractive index

n20/D 1.485 (lit.)

useful pH range

7.3-8.3

pKa (25 °C)

7.8

bp

190-193 °C/5 mmHg (lit.)

mp

17.9-21 °C (lit.)

density

1.124 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

OCCN(CCO)CCO

InChI

1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2

InChI key

GSEJCLTVZPLZKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

354.2 °F - closed cup

flash_point_c

179 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoxuan Liu et al.
Molecular pharmaceutics, 9(3), 470-481 (2012-01-03)
Successful achievement of RNA interference in therapeutic applications requires safe and efficient vectors for siRNA delivery. In the present study, we demonstrate that a triethanolamine (TEA)-core PAMAM dendrimer of generation 5 (G(5)) is able to deliver sticky siRNAs bearing complementary
Stuart K Langley et al.
Dalton transactions (Cambridge, England : 2003), 41(32), 9789-9796 (2012-07-11)
The synthesis, structures and magnetic properties of two new mixed-valence heptanuclear manganese clusters are described. Both complexes utilize triethanolamine (teaH(3)) as a bridging ligand, displaying near planar, disc-like metal topologies and are of formulae [Mn(II)(4)Mn(IV)(3)(tea)(teaH(2))(3)(peolH)(4)](BF(4))(2)·solv (1) and [Mn(II)(4)Mn(III)(3)F(3)(tea)(teaH)(teaH(2))(2)(piv)(4)(Hpiv)(chp)(3)]·0.5MeCN (2). Compound
Xiaoxuan Liu et al.
Bioconjugate chemistry, 22(12), 2461-2473 (2011-11-08)
With the aim of developing dendrimer nanovectors with a precisely controlled architecture and flexible structure for DNA transfection, we designed PAMAM dendrimers bearing a triethanolamine (TEA) core, with branching units pointing away from the center to create void spaces, reduce
Liviu Ungur et al.
Journal of the American Chemical Society, 134(45), 18554-18557 (2012-11-01)
A toroidal magnetic moment in the absence of conventional total magnetic moment is observed in a {Dy(6)} ring. The reason for the net toroidal arrangement of the local magnetic moments is the high symmetry of the complex in combination with
Kostas Karatasos et al.
Macromolecular bioscience, 12(2), 225-240 (2011-12-08)
In this work we report, compare and discuss the results obtained from fully atomistic molecular dynamics simulations of generations 4, 5, and 6 of PAMAM-based dendrimers having NH(3) and triethanolamine as cores, forming complexes with a short interfering RNA (siRNA)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service