Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800435

Sigma-Aldrich

Sodium citrate tribasic dihydrate

LR, ≥99%

Synonym(s):

Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10
Beilstein/REAXYS Number:
6104939
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
assay:
≥99%
grade:
LR
form:
crystalline powder
Pricing and availability is not currently available.

grade

LR

product line

Vetec

assay

≥99%

form

crystalline powder

pH

7.5-9.0 (5% in water)

mp

>300 °C (lit.)

SMILES string

O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3

InChI key

NLJMYIDDQXHKNR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evgeny Smirnov et al.
ACS nano, 9(6), 6565-6575 (2015-06-04)
Soft or "liquid-liquid" interfaces were functionalized by roughly half a monolayer of mirror-like nanofilms of gold nanoparticles using a precise interfacial microinjection method. The surface coverage of the nanofilm was characterized by ion transfer voltammetry. These gold nanoparticle films represent
Jun Zhao et al.
Nanotechnology, 25(40), 405101-405101 (2014-09-12)
The treatment of liver injuries or hepatocellular carcinoma (HCC) has been hindered by the lack of efficient drug delivery. Even with the help of nanoparticles or other synthetic delivering agents, a large portion of the dose is still sequestered in
Ruben de Vries et al.
Journal of dairy science, 98(12), 8678-8687 (2015-10-12)
The aim of this study was to evaluate the effect of shortening or omitting the dry period of dairy cows on milk casein composition. For this study, we analyzed milk samples of 90 cows with a dry period of 0
Stijn Mertens et al.
Applied and environmental microbiology, 81(23), 8202-8214 (2015-09-27)
Lager beer is the most consumed alcoholic beverage in the world. Its production process is marked by a fermentation conducted at low (8 to 15°C) temperatures and by the use of Saccharomyces pastorianus, an interspecific hybrid between Saccharomyces cerevisiae and
Lakshminarayana Polavarapu et al.
Small (Weinheim an der Bergstrasse, Germany), 10(15), 3065-3071 (2014-05-03)
The translation of a technology from the laboratory into the real world should meet the demand of economic viability and operational simplicity. Inspired by recent advances in conductive ink pens for electronic devices on paper, we present a "pen-on-paper" approach

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service