V800425
Thiourea
LR, ≥98%
Synonym(s):
Sulfourea, Thiocarbamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH2CSNH2
CAS Number:
Molecular Weight:
76.12
Beilstein/REAXYS Number:
605327
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Recommended Products
grade
LR
product line
Vetec™
assay
≥98%
mp
170-176 °C (lit.)
SMILES string
NC(N)=S
InChI
1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)
InChI key
UMGDCJDMYOKAJW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Chaotropic agent; strong denaturant. Increases solubility and recovery of proteins
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Elaine M Boyle et al.
The Journal of organic chemistry, 78(17), 8312-8319 (2013-08-01)
Thiourea-functionalized Tröger's base receptors 1 and 2 have been synthesized and evaluated as novel for the recognition of anions. Receptor 2 gave rise to significant changes in the absorption spectrum upon titration with AcO(-) and H2PO4(-) and acted as a
David Weisbrod et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(18), E1685-E1694 (2013-04-17)
Proper expression and function of the cardiac pacemaker is a critical feature of heart physiology. Two main mechanisms have been proposed: (i) the "voltage-clock," where the hyperpolarization-activated funny current If causes diastolic depolarization that triggers action potential cycling; and (ii)
Mireille Vonlanthen et al.
The Journal of organic chemistry, 78(8), 3980-3988 (2013-03-09)
Proof that sulfur is a viable reporting element for the development of fluorescent chemosensors for metal ions is presented. To date, the majority of metal-responsive fluorescent chemosensors have relied on metal-nitrogen coordination to provide a fluorescence response, most commonly by
Mathieu P Lalonde et al.
Journal of the American Chemical Society, 135(5), 1891-1894 (2013-01-17)
A highly enantio- and diastereoselective synthesis of indolo- and benzoquinolizidine compounds has been developed through the formal aza-Diels-Alder reaction of enones with cyclic imines. This transformation is catalyzed by a new bifunctional primary aminothiourea that achieves simultaneous activation of both
Shrinivas Venkataraman et al.
Macromolecular rapid communications, 34(8), 652-658 (2013-03-14)
Readily water-soluble PEGylated amphiphiles containing bis-thiourea-based molecular recognition units at the interface of hydrophobic and hydrophilic blocks are developed. Self-assembly of these amphiphiles is found to be dependent on the exact chemical composition of the hydrophobic component. Elongated, spherical, and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service