Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800415

Sigma-Aldrich

D-(−)-Tartaric acid

LR, ≥98%

Synonym(s):

(2S,3S)-(−)-Tartaric acid, D-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
Beilstein/REAXYS Number:
1725145
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

LR

description

synthetic

product line

Vetec

assay

≥98%

mp

172-174 °C (lit.)

SMILES string

O[C@@H]([C@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m0/s1

InChI key

FEWJPZIEWOKRBE-LWMBPPNESA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Unnatural isomer

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Bencini et al.
Chemical communications (Cambridge, England), 48(84), 10428-10430 (2012-09-18)
A chiral ditopic polyammonium receptor featuring two [9]aneN(3) moieties separated by a (S)-BINOL linker is able to selectively bind and sense in water (S,S)-tartaric acid over its (R,R)/meso forms.
Carmen-Alice Teacă et al.
Carbohydrate polymers, 93(1), 307-315 (2013-03-08)
The present paper describes the preparation and characterization of polysaccharides-based bio-composite films obtained by the incorporation of 10, 20 and 30 wt% birch cellulose (BC) within a glycerol plasticized matrix constituted by the corn starch (S) and chemical modified starch
Anne Toboldt et al.
Applied and environmental microbiology, 78(20), 7347-7357 (2012-08-14)
In this study, the population structure, incidence, and potential sources of human infection caused by the d-tartrate-fermenting variant of Salmonella enterica serovar Paratyphi B [S. Paratyphi B (dT+)] was investigated. In Germany, the serovar is frequently isolated from broilers. Therefore
J W H Smith et al.
Journal of hazardous materials, 235-236, 279-285 (2012-09-07)
Impregnated activated carbons (IACs) that are used in multi-gas respirator applications usually contain copper and/or zinc impregnants. Co-impregnating with properly selected acids can improve the distribution of the metallic impregnant on the carbon and improve the gas adsorption capacity of
Prashant K Metri et al.
Chemistry, an Asian journal, 8(2), 488-493 (2012-12-05)
Total synthesis of the polyhydroxy caprolactam amide natural product, bengamide E, is accomplished starting from tartaric acid. Key reactions in the synthesis include desymmetrization of the bis(dimethylamide) unit of tartaric acid, Zn(BH(4))(2)-mediated anti-selective reduction, and a Horner-Wadsworth-Emmons olefination.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service