Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800411

Sigma-Aldrich

Sulfur

LR, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
EC Number:
MDL number:
UNSPSC Code:
12141912
PubChem Substance ID:
assay:
≥99%
grade:
LR
form:
solid

grade

LR

vapor density

8.9 (vs air)

vapor pressure

1 mmHg ( 183.8 °C)
10 mmHg ( 246 °C)

product line

Vetec

assay

≥99%

form

solid

autoignition temp.

450 °F

resistivity

2E23 μΩ-cm, 20°C

bp

444.7 °C (lit.)

mp

112.8 °C (rhombic) (lit.)
117-120 °C (lit.)
119.0 °C (monoclinic) (lit.)

SMILES string

[S]

InChI

1S/S

InChI key

NINIDFKCEFEMDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farhad Forouhar et al.
Nature chemical biology, 9(5), 333-338 (2013-04-02)
How living organisms create carbon-sulfur bonds during the biosynthesis of critical sulfur-containing compounds is still poorly understood. The methylthiotransferases MiaB and RimO catalyze sulfur insertion into tRNAs and ribosomal protein S12, respectively. Both belong to a subgroup of radical-S-adenosylmethionine (radical-SAM)
Mark A Lever et al.
Science (New York, N.Y.), 339(6125), 1305-1308 (2013-03-16)
Sediment-covered basalt on the flanks of mid-ocean ridges constitutes most of Earth's oceanic crust, but the composition and metabolic function of its microbial ecosystem are largely unknown. By drilling into 3.5-million-year-old subseafloor basalt, we demonstrated the presence of methane- and
William Tam et al.
Journal of molecular biology, 425(14), 2450-2462 (2013-04-02)
The assembly of long non-contractile phage tails begins with the formation of the tail tip complex (TTC). TTCs are multi-functional protein structures that mediate host cell adsorption and genome injection. The TTC of phage λ is assembled from multiple copies
Gulluzar Bastug et al.
The Journal of organic chemistry, 78(18), 9303-9308 (2013-08-14)
The newly prepared complex [Pd(IPr*(OMe))(cin)(Cl)] provides high catalytic activity for carbon-sulfur cross-coupling reactions. Nonactivated and deactivated aryl halides were successfully coupled with a large variety of aryl- and alkylthiols using this well-defined palladium N-heterocyclic carbene (NHC) complex.
Chemoselective peptide ligation-desulfurization at aspartate.
Robert E Thompson et al.
Angewandte Chemie (International ed. in English), 52(37), 9723-9727 (2013-07-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service