Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800409

Sigma-Aldrich

Sucrose

AR

Synonym(s):

α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein/REAXYS Number:
90825
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Pricing and availability is not currently available.

grade

AR

product line

Vetec

assay

≥99.5%

mp

185-187 °C (lit.)

SMILES string

OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

CZMRCDWAGMRECN-UGDNZRGBSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean Grisouard et al.
Blood, 125(13), 2131-2140 (2015-01-18)
The acquired somatic JAK2-V617F mutation is present in >80% of patients with myeloproliferative neoplasms (MPNs). Stat3 plays a role in hematopoietic homeostasis and might influence the JAK2-V617F-driven MPN phenotype. We crossed our transgenic SclCre;V617F mice with a conditional Stat3 knockout
Marcus Karlstetter et al.
Progress in retinal and eye research, 45, 30-57 (2014-12-06)
Resident microglial cells can be regarded as the immunological watchdogs of the brain and the retina. They are active sensors of their neuronal microenvironment and rapidly respond to various insults with a morphological and functional transformation into reactive phagocytes. There
William J Armour et al.
The Plant cell, 27(9), 2484-2500 (2015-08-25)
Lobe development in the epidermal pavement cells of Arabidopsis thaliana cotyledons and leaves is thought to take place via tip-like growth on the concave side of lobes driven by localized concentrations of actin filaments and associated proteins, with a predicted
Akie Shimotohno et al.
Plant & cell physiology, 56(4), 620-630 (2015-02-12)
Boron, an essential micronutrient, is transported in roots of Arabidopsis thaliana mainly by two different types of transporters, BORs and NIPs (nodulin26-like intrinsic proteins). Both are plasma membrane localized, but have distinct transport properties and patterns of cell type-specific accumulation
Julia B Cordero et al.
The EMBO journal, 33(13), 1474-1491 (2014-05-03)
The non-receptor tyrosine kinase c-Src, hereafter referred to as Src, is overexpressed or activated in multiple human malignancies. There has been much speculation about the functional role of Src in colorectal cancer (CRC), with Src amplification and potential activating mutations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service