Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800408

Sigma-Aldrich

Starch

LR, from potato, solubility:, soluble

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
Pricing and availability is not currently available.

biological source

potato

grade

LR

product line

Vetec

solubility

soluble

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Wu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 928, 106-112 (2013-04-23)
The aqueous two-phase system (ATPS) with polyethylene glycol (PEG) and hydroxypropyl starch (HPS) was evaluated for the extraction of human immunoglobulin G (IgG) from human serum albumin (HSA). The partitions of IgG in PEG/sulphate, PEG/phosphate, PEG/Dextran and PEG/HPS ATPSs were
Reply to J.E. Galgani and G. Valentino.
Coby Eelderink et al.
The American journal of clinical nutrition, 97(4), 903-904 (2013-05-01)
Li Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(14), 5380-5385 (2013-03-20)
Three grinding stones from Shizitan Locality 14 (ca. 23,000-19,500 calendar years before present) in the middle Yellow River region were subjected to usewear and residue analyses to investigate human adaptation during the last glacial maximum (LGM) period, when resources were
Ayse Bengisu Seferoglu et al.
Plant science : an international journal of experimental plant biology, 205-206, 29-37 (2013-03-19)
ADP-glucose pyrophosphorylase (AGPase) is a key enzyme in plant starch biosynthesis. It contains large (LS) and small (SS) subunits encoded by two different genes. In this study, we explored the transcriptional regulation of both the LS and SS subunits of
FDA delays decision to remove powdered gloves.
Richard F Edlich et al.
JAMA surgery, 148(5), 406-407 (2013-05-17)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service