Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800387

Sigma-Aldrich

Sodium dodecyl sulfate

LR, ≥90%

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein/REAXYS Number:
3599286
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
Pricing and availability is not currently available.

grade

LR

description

anionic detergent

product line

Vetec

assay

≥90%

mol wt

288.38 g/mol

mp

204-207 °C (lit.)

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

application

Anionic detergent

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

338.0 °F

flash_point_c

170 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Maillard et al.
Molecular psychiatry, 20(1), 140-147 (2014-11-26)
Anatomical structures and mechanisms linking genes to neuropsychiatric disorders are not deciphered. Reciprocal copy number variants at the 16p11.2 BP4-BP5 locus offer a unique opportunity to study the intermediate phenotypes in carriers at high risk for autism spectrum disorder (ASD)
Shane Stegeman et al.
Cancer discovery, 5(4), 368-379 (2015-02-19)
Prostate cancer is the second most common malignancy among men worldwide. Genome-wide association studies have identified 100 risk variants for prostate cancer, which can explain approximately 33% of the familial risk of the disease. We hypothesized that a comprehensive analysis
S Desrivières et al.
Molecular psychiatry, 20(2), 263-274 (2014-02-12)
Despite the recognition that cortical thickness is heritable and correlates with intellectual ability in children and adolescents, the genes contributing to individual differences in these traits remain unknown. We conducted a large-scale association study in 1583 adolescents to identify genes
Susanne Kohl et al.
Nature genetics, 47(7), 757-765 (2015-06-02)
Achromatopsia (ACHM) is an autosomal recessive disorder characterized by color blindness, photophobia, nystagmus and severely reduced visual acuity. Using homozygosity mapping and whole-exome and candidate gene sequencing, we identified ten families carrying six homozygous and two compound-heterozygous mutations in the
Yan Xiang et al.
The Journal of clinical investigation, 125(6), 2293-2306 (2015-04-29)
Glutaminase (GLS), which converts glutamine to glutamate, plays a key role in cancer cell metabolism, growth, and proliferation. GLS is being explored as a cancer therapeutic target, but whether GLS inhibitors affect cancer cell-autonomous growth or the host microenvironment or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service