Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V800386

Sigma-Aldrich

Sodium dodecyl sulfate

AR, ≥99%

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein/REAXYS Number:
3599286
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
Pricing and availability is not currently available.

grade

AR

description

anionic detergent

product line

Vetec

assay

≥99%

mol wt

288.38 g/mol

mp

204-207 °C (lit.)

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Anionic detergent

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

338.0 °F

flash_point_c

170 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Tian et al.
Disease models & mechanisms, 8(6), 635-641 (2015-06-04)
Nemaline myopathy (NM) is one of the most common forms of congenital myopathy, and affects either fast myofibers, slow myofibers, or both. However, an animal model for congenital myopathy with fast-myofiber-specific atrophy is not available. Furthermore, mutations in the leiomodin-3
Lan Zhang et al.
BMC cancer, 14, 885-885 (2014-11-28)
Upregulator of cell proliferation 4 (URG4) has been implicated in the oncogenesis of certain cancers. However, the correlation between URG4 expression and clinicopathological significance in human cancer remains unclear. Therefore, this study investigated its expression and clinicopathological significance in cervical
Carolina Ceballos-Diaz et al.
Molecular neurodegeneration, 10, 25-25 (2015-07-15)
UBQLN2 mutations have recently been associated with familial forms of amyotrophic lateral sclerosis (ALS) and ALS-dementia. UBQLN2 encodes for ubiquilin-2, a member of the ubiquitin-like protein family which facilitates delivery of ubiquitinated proteins to the proteasome for degradation. To study
Paolo Ceppi et al.
Nature communications, 5, 5238-5238 (2014-11-05)
CD95 (APO-1/Fas) is a death receptor used by immune cells to kill cancer cells through induction of apoptosis. However, the elimination of CD95 or its ligand, CD95L, from cancer cells results in death induced by CD95R/L elimination (DICE), a type
Jennifer Hirst et al.
Human molecular genetics, 24(17), 4984-4996 (2015-06-19)
Adaptor proteins (AP 1-5) are heterotetrameric complexes that facilitate specialized cargo sorting in vesicular-mediated trafficking. Mutations in AP5Z1, encoding a subunit of the AP-5 complex, have been reported to cause hereditary spastic paraplegia (HSP), although their impact at the cellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service