Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800368

Sigma-Aldrich

Sodium bicarbonate

LR, anhydrous, ≥99%

Synonym(s):

Sodium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHCO3
CAS Number:
Molecular Weight:
84.01
Beilstein/REAXYS Number:
4153970
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99%
grade:
LR
anhydrous
form:
crystalline powder
Pricing and availability is not currently available.

grade

LR
anhydrous

product line

Vetec

assay

≥99%

form

crystalline powder

pH

8.0-8.5 (1% in water)

pKa (25 °C)

(1) 6.37, (2) 10.25 (carbonic acid)

density

2.16 g/mL at 25 °C (lit.)

SMILES string

[Na+].OC([O-])=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

UIIMBOGNXHQVGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium bicarbonate (NaHCO3), generally known as baking powder, is commonly used as a base in many organic reactions. Recently, it has also been used in organic electronic devices as an electron injection layer material.[1][2]

Application

Sodium bicarbonate can be used:
  • As a catalyst or a promoter in the acetylation of phenols and primary alcohols using acetic anhydride.[3]
  • In the synthesis of sugar orthoesters.[4]
  • In the asymmetric isomerization studies of cyclic dienes.[5]

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile and efficient acetylation of primary alcohols and phenols with acetic anhydride catalyzed by dried sodium bicarbonate
Lugemwa F, et al.
Catalysts, 3(4), 954-965 (2013)
Baking soda: an ultra-cheap and air stable electron injection layer for organic electronic devices
Long D and Noh Y
Journal of Material Chemistry C, 6(47), 12871-12878 (2018)
Asymmetric Isomerisation of a Cyclic Diene: a Comparative Study of BINAP and BIPNOR-Rhodium (I) Catalysts
Faitg T, et al.
Tetrahedron, 56(1), 101-104 (2000)
Baking soda
Sodium bicarbonate (2013)
A facile method for the preparation of sugar orthoesters promoted by anhydrous sodium bicarbonate
Wei S, et al.
Canadian Journal of Chemistry, 87(12), 1733-1737 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service