Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800347

Sigma-Aldrich

Pyridine

AR, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
Beilstein/REAXYS Number:
103233
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
grade:
AR
assay:
≥99.5%
bp:
115 °C (lit.)
vapor pressure:
10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

AR

vapor density

2.72 (vs air)

vapor pressure

10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

product line

Vetec

assay

≥99.5%

form

liquid

autoignition temp.

899 °F

expl. lim.

12.4 %

dilution

(for analytical testing)

refractive index

n20/D 1.509 (lit.)

pH

8.5 (25 °C, 15.82 g/L)

bp

115 °C (lit.)

mp

−42 °C (lit.)

density

0.978 g/mL at 25 °C (lit.)

SMILES string

C1=CN=CC=C1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

68.0 °F - closed cup

flash_point_c

20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total syntheses of complanadines A and B.
Le Zhao et al.
Angewandte Chemie (International ed. in English), 52(6), 1722-1725 (2013-01-03)
Maria Milina et al.
Angewandte Chemie (International ed. in English), 54(5), 1591-1594 (2014-11-08)
Without techniques sensitive to complex pore architectures, synthetic efforts to enhance molecular transport in zeolite and other porous materials through hierarchical structuring lack descriptors for their rational design. The power of positron annihilation lifetime spectroscopy (PALS) to characterize the pore
Nathalie Ségaud et al.
Inorganic chemistry, 52(2), 691-700 (2013-01-11)
We report the synthesis, characterization, and solution chemistry of a series of new Fe(II) complexes based on the tetradentate ligand N-methyl-N,N'-bis(2-pyridyl-methyl)-1,2-diaminoethane or the pentadentate ones N,N',N'-tris(2-pyridyl-methyl)-1,2-diaminoethane and N,N',N'-tris(2-pyridyl-methyl)-1,3-diaminopropane, modified by propynyl or methoxyphenyltriazolyl groups on the amino functions. Six of
Ye Wei et al.
Journal of the American Chemical Society, 135(10), 3756-3759 (2013-02-27)
We describe here a [3+3]-type condensation reaction of O-acetyl ketoximes and α,β-unsaturated aldehydes that is synergistically catalyzed by a copper(I) salt and a secondary ammonium salt (or amine). This redox-neutral reaction allows modular synthesis of a variety of substituted pyridines
Jerri M Rook et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(3), 755-765 (2014-09-23)
Allosteric modulators of the metabotropic glutamate receptor subtype 5 (mGlu5) have exciting potential as therapeutic agents for multiple brain disorders. Translational studies with mGlu5 modulators have relied on mGlu5 allosteric site positron emission tomography (PET) radioligands to assess receptor occupancy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service