Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800313

Sigma-Aldrich

Potassium chromate

LR, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CrO4
CAS Number:
Molecular Weight:
194.19
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
assay:
≥99%
grade:
LR
form:
solid
Pricing and availability is not currently available.

grade

LR

vapor density

6.7 (vs air)

product line

Vetec

assay

≥99%

form

solid

pH

8.5-10.0 (20 °C, 50 g/L)

mp

971 °C (lit.)

SMILES string

[K+].[K+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.2K.4O/q;2*+1;;;2*-1

InChI key

XMXNVYPJWBTAHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saturnino Spiga et al.
Brain structure & function, 216(3), 171-182 (2011-04-05)
Visualization of neuronal elements is of fundamental importance in modern neuroscience. Golgi-Cox impregnation is a widely employed method that provides detailed information about morphological characteristics of neurons, but none regarding their neurochemical features. Immunocytochemical procedures, on the other hand, can
Xiao-Zhang Yu et al.
Ecotoxicology (London, England), 19(1), 61-68 (2009-09-04)
The removal of hexavalent and trivalent chromium from hydroponic solution by plants to changes in temperature was investigated. Pre-rooted hybrid willows (Salix matsudana Koidz x alba L.) were exposed to a nutrient solution spiked with potassium chromate (K(2)CrO(4)) or chromium
Ying Li et al.
Colloids and surfaces. B, Biointerfaces, 65(1), 25-29 (2008-04-02)
Adsorption thermodynamic characteristic and kinetics of dissolved chromium from aqueous solutions onto humic acids (HAs) were studied. The results showed that the Freundlich isotherm model described the equilibrium data well. The negative DeltaG degrees confirmed the spontaneity of adsorption process.
Nejla Soudani et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 63(6), 541-548 (2010-05-25)
Chromium, a major environmental pollutant, is known for its wide toxic manifestations. The present experiment pertains to the protective role of selenium (Se) against K(2)Cr(2)O(7)-induced hepatotoxicity. Female Wistar rats were divided into four groups of six each: group I served
Inês Domingues et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 152(3), 338-345 (2010-06-23)
The present work aims to compare the sensitivity of embryos and adult zebrafish to chromium (VI) (as potassium dichromate) focusing on biomarkers (cholinesterase, glutathione S-transferase and lactate dehydrogenase) as endpoints. Zebrafish eggs showed less sensitivity to Cr (VI) (96 h-LC50=145.7

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service