Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800294

Sigma-Aldrich

Perchloric acid solution

0.1 N in acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

product line

Vetec

form

liquid

concentration

0.1 N in acetic acid

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Perchloric acid (in acetic acid) can be used as a detosylating agent for the deprotection of the tosyl functional groups in p-methyl benzene sulfonyl derivatives of primary, secondary aryl amines, and chiral aminoketones.[1] It can also be used in the aromatic mercuration reaction.[2]

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Flame over circle

signalword

Danger

hcodes

pcodes

Hazard Classifications

Ox. Liq. 2

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Perchloric Acid catalyzed aromatic mercuration in acetic acid solution. II. The Substitution Process
Kresge, A and Brennan, J
The Journal of Organic Chemistry, 32(3), 752-755 (1967)
Perchloric Acid-Acetic Acid: A Reagent System for Detosylation
Kudav, DP, et al.
Synthetic Communications, 17(10), 1185-1187 (1987)
John Greenwood et al.
ACS nano, 9(5), 5520-5535 (2015-04-22)
We shine light on the covalent modification of graphite and graphene substrates using diazonium chemistry under ambient conditions. We report on the nature of the chemical modification of these graphitic substrates, the relation between molecular structure and film morphology, and
Martin G Liebensteiner et al.
Science (New York, N.Y.), 340(6128), 85-87 (2013-04-06)
Perchlorate and chlorate anions [(per)chlorate] exist in the environment from natural and anthropogenic sources, where they can serve as electron acceptors for bacteria. We performed growth experiments combined with genomic and proteomic analyses of the hyperthermophile Archaeoglobus fulgidus that show
Dipanwita Das et al.
Journal of the American Chemical Society, 135(10), 4018-4026 (2013-02-28)
Catalytic four-electron reduction of O2 by ferrocene (Fc) and 1,1'-dimethylferrocene (Me2Fc) occurs efficiently with a dinuclear copper(II) complex [Cu(II)2(XYLO)(OH)](2+) (1), where XYLO is a m-xylene-linked bis[(2-(2-pyridyl)ethyl)amine] dinucleating ligand with copper-bridging phenolate moiety], in the presence of perchloric acid (HClO4) in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service