Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800283

Sigma-Aldrich

Nitrobenzene

LR, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein/REAXYS Number:
507540
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
grade:
LR
assay:
≥99%
bp:
210-211 °C (lit.)
vapor pressure:
0.15 mmHg ( 20 °C)
50 mmHg ( 120 °C)
Pricing and availability is not currently available.

grade

LR

vapor density

4.2 (vs air)

vapor pressure

0.15 mmHg ( 20 °C)
50 mmHg ( 120 °C)

product line

Vetec

assay

≥99%

form

liquid

autoignition temp.

899 °F

expl. lim.

40 %

dilution

(for analytical testing)

refractive index

n20/D 1.551 (lit.)

pH

8-8.5 (20 °C, 1 g/L)

bp

210-211 °C (lit.)

mp

5-6 °C (lit.)

density

1.196 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccccc1

InChI

1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H

InChI key

LQNUZADURLCDLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Repr. 1B - STOT RE 1 Inhalation

target_organs

Blood

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

190.4 °F - closed cup

flash_point_c

88 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Emmanuel et al.
Journal of hazardous materials, 279, 117-124 (2014-07-23)
The present study involves a green synthesis of gold nanoparticles (Au-NPs) using Acacia nilotica twig bark extract at room temperature and trace level detection of one of the hazardous materials, viz. nitrobenzene (NB) that causes Methemoglobinaemia. The synthesis protocol demonstrates
Jiping Jiang et al.
Journal of hazardous materials, 227-228, 280-291 (2012-06-06)
This paper presents a generic framework and decision tools of real-time risk assessment on Emergency Environmental Decision Support System for response to chemical spills in river basin. The generic "4-step-3-model" framework is able to delineate the warning area and the
Hao Wu et al.
Chemical communications (Cambridge, England), 49(34), 3516-3518 (2013-02-12)
A photo-, acid- and thermo-responsive nitrobenzene-based amphiphilic copolymer was synthesized and the corresponding multi-responsive behavior of the copolymer assemblies was revealed by TEM, DLS and AFM. The morphological changes of the assemblies under multi-stimuli endowed them with a controlled release
Wataru Kosaka et al.
Chemical communications (Cambridge, England), 49(16), 1594-1596 (2012-11-24)
A porous one-dimensional coordination chain compound, [Ru(2)(p-F-PhCO(2))(4)(phz)] (; p-F-PhCO(2)(-) = para-fluorobenzoate; phz = phenazine) derived by drying its nitrobenzene-solvated compound, specifically adsorbs CO(2) at 195 K in a stepwise sorption manner.
Yingzi Lin et al.
Bioresource technology, 118, 128-135 (2012-06-19)
A process combining an Anaerobic Baffled Reactor (ABR) and a constructed wetland was employed to treat nitrobenzene wastewater. The overall performance was examined throughout long-term operation with a hydraulic retention time (HRT) of 24 h at 30±1 °C. The effluent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service