Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800279

Sigma-Aldrich

Nickel(II) sulfate hexahydrate

LR, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiSO4 · 6H2O
CAS Number:
Molecular Weight:
262.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥98%
grade:
LR
form:
crystalline powder
Pricing and availability is not currently available.

grade

LR

vapor density

2.07 (vs air)

product line

Vetec

assay

≥98%

form

crystalline powder

pH

4.5

SMILES string

O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

InChI

1S/Ni.H2O4S.6H2O/c;1-5(2,3)4;;;;;;/h;(H2,1,2,3,4);6*1H2/q+2;;;;;;;/p-2

InChI key

RRIWRJBSCGCBID-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atharva Sahasrabudhe et al.
Nature communications, 9(1), 2014-2014 (2018-05-24)
Herein, we present an innovative approach for transforming commonly available cellulose paper into a flexible and catalytic current collector for overall water splitting. A solution processed soak-and-coat method of electroless plating was used to render a piece of paper conducting
Monica Mattarozzi et al.
Micron (Oxford, England : 1993), 75, 18-26 (2015-05-20)
Environmental scanning electron microscopy (ESEM) permits to analyze samples in their native-hydrated state, allowing a broad spectrum of biological applications. In this study, ESEM equipped with energy dispersive X-ray spectrometer (EDS) was used as a fast method to analyze tissue
K Amudha et al.
Chemico-biological interactions, 193(1), 57-64 (2011-05-24)
This study was conducted to investigate the beneficial role of naringin on nickel induced nephrotoxicity. Nickel (Ni) (20mg/kg body weight (b.w.) was administered intraperitoneally (i.p.) for 20 days. Naringin was administered orally (20, 40 and 80 mg/kg b.w.) with i.p.
Xiaohua Xu et al.
Particle and fibre toxicology, 9, 40-40 (2012-11-07)
It has been well recognized that toxicity of fine ambient air particulate matter (PM(2.5)) may depend on its chemical constituents, including components such as soluble metals that may theoretically exert distinctive effects. We have recently demonstrated an important effect of
A Tammaro et al.
International journal of immunopathology and pharmacology, 25(2), 519-521 (2012-06-16)
Nickel deficiency leads to reduced iron content in organs and to reduced hemoglobin levels and haematocrit counts. We carried out a clinical trial of oral hyposensitization therapy with low doses of nickel on a group of 20 patients affected by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service