Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V800278

Sigma-Aldrich

Nickel(II) chloride

LR, 44.6-46% Ni basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiCl2
CAS Number:
Molecular Weight:
129.60
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
powder or chunks
Pricing and availability is not currently available.

grade

LR

product line

Vetec

form

powder or chunks

concentration

44.6-46% Ni

density

3.55 g/mL at 25 °C (lit.)

SMILES string

Cl[Ni]Cl

InChI

1S/2ClH.Ni/h2*1H;/q;;+2/p-2

InChI key

QMMRZOWCJAIUJA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

The vapor-phase co-reductions with other metal halides such as aluminum chloride (cat. no. 449598) by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamzin A Blewett et al.
Archives of environmental contamination and toxicology, 68(2), 382-394 (2014-12-30)
The mechanisms of nickel (Ni) toxicity in marine fish remain unclear, although evidence from freshwater (FW) fish suggests that Ni can act as a pro-oxidant. This study investigated the oxidative stress effects of Ni on the euryhaline killifish (Fundulus heteroclitus)
Katherine R Brimblecombe et al.
ACS chemical neuroscience, 6(1), 124-129 (2014-12-02)
Neuronal T-type voltage-gated Ca(2+) channels are reported to have physiological roles that include regulation of burst firing, Ca(2+) oscillations, and neurotransmitter release. These roles are often exposed experimentally by blocking T-type channels with micromolar Ni(2+). We used Ni(2+) to explore
John M Besser et al.
Environmental toxicology and chemistry, 32(11), 2495-2506 (2013-05-10)
This study evaluated the chronic toxicity of Ni-spiked freshwater sediments to benthic invertebrates. A 2-step spiking procedure (spiking and sediment dilution) and a 2-stage equilibration period (10 wk anaerobic and 1 wk aerobic) were used to spike 8 freshwater sediments with wide
William G Brumbaugh et al.
Environmental toxicology and chemistry, 32(11), 2482-2494 (2013-05-10)
Two spiking methods were compared and nickel (Ni) partitioning was evaluated during a series of toxicity tests with 8 different freshwater sediments having a range of physicochemical characteristics. A 2-step spiking approach with immediate pH adjustment by addition of NaOH
M Song et al.
British journal of pharmacology, 171(14), 3432-3447 (2014-03-20)
SKF 96365 is well known for its suppressing effect on human glioblastoma growth by inhibiting pre-activated transient receptor potential canonical (TRPC) channels and Ca(2+) influx. The effect of SKF 96363 on glioblastoma cells, however, may be multifaceted and this possibility

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service