Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800267

Sigma-Aldrich

Heptane

suitable for HPLC, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
Beilstein/REAXYS Number:
1730763
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
HPLC grade
assay:
≥99.5%
technique(s):
HPLC: suitable
bp:
98 °C (lit.)
vapor pressure:
40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)
Pricing and availability is not currently available.

grade

HPLC grade

Quality Level

vapor density

3.5 (vs air)

vapor pressure

40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

product line

Vetec

assay

≥99.5%

form

liquid

autoignition temp.

433 °F

expl. lim.

7 %

technique(s)

HPLC: suitable

refractive index

n20/D 1.387 (lit.)

bp

98 °C (lit.)

mp

−91 °C (lit.)

density

0.684 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

24.8 °F - closed cup

flash_point_c

-4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chris G C Jacobs et al.
Insect biochemistry and molecular biology, 60, 7-12 (2015-03-10)
Insects have been extraordinary successful in colonizing terrestrial habitats and this success is partly due to a protective cuticle that mainly contains chitin and proteins. The cuticle has been well studied in larvae and adults, but little attention has been
Arun K Kota et al.
Advanced materials (Deerfield Beach, Fla.), 24(43), 5838-5843 (2012-08-30)
Hierarchically structured, superoleophobic surfaces are demonstrated that display one of the lowest contact angle hysteresis values ever reported - even with extremely low-surface-tension liquids such as n-heptane. Consequently, these surfaces allow, for the first time, even ≈2 μL n-heptane droplets
Bing Chen et al.
Journal of hazardous materials, 188(1-3), 369-374 (2011-02-26)
The influence of the initial fuel temperature on the burning behavior of n-heptane pool fire was experimentally studied at the State Key Laboratory of Fire Science (SKLFS) large test hall. Circular pool fires with diameters of 100mm, 141 mm, and
Thushara Karunasekara et al.
Journal of chromatography. A, 1218(6), 809-816 (2011-01-07)
Partition coefficients for varied compounds were determined for the organic solvent-propylene carbonate biphasic partition system where the organic solvent is n-heptane, isopentyl ether or 1-octanol. These partition coefficient databases are analyzed using the solvation parameter model facilitating a quantitative comparison
Manuela Nagel et al.
Plant, cell & environment, 38(6), 1011-1022 (2014-10-21)
Globally, over 7.4 million accessions of crop seeds are stored in gene banks, and conservation of genotypic variation is pivotal for breeding. We combined genetic and biochemical approaches to obtain a broad overview of factors that influence seed storability and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service