Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800256

Sigma-Aldrich

Mercury(II) sulfate

LR, ≥99%

Synonym(s):

Mercuric sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgSO4
CAS Number:
Molecular Weight:
296.65
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99%
grade:
LR
form:
crystalline powder
Pricing and availability is not currently available.

grade

LR

product line

Vetec

assay

≥99%

form

crystalline powder

SMILES string

[Hg++].[O-]S([O-])(=O)=O

InChI

1S/Hg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

DOBUSJIVSSJEDA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

target_organs

Kidney

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shovana Pradhan et al.
Chemosphere, 136, 198-203 (2015-05-24)
Reverse osmosis (RO) concentrate (ROC) streams generated from RO-based municipal wastewater reclamation processes pose potential health and environmental risks on their disposal to confined water bodies such as bays. A UV/H2O2 advanced oxidation process followed by a biological activated carbon
Ai-hua Liang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 30(4), 249-252 (2005-02-24)
Traditional Chinese medicine (TCM) experienced a gradual course in recognition of the toxicity of Cinnabaris from "nontoxic" to "toxic". The ancient doctors of TCM understood both the toxic property and the regularity of increasing toxicity of Cinnabaris. In long-term clinical
[A case of granulomatous reaction to tattooing].
R Colombo et al.
Minerva chirurgica, 42(8), 747-749 (1987-04-30)
B Mora et al.
Canadian journal of microbiology, 26(8), 930-937 (1980-08-01)
Studies have been made on the toxicities of three inorganic (HgCl2, HgSO4, and NH2HgCl) and two organic (CH3HgCl and phenyl mercuric acetate (PMA)) mercury derivatives on planktonic algae (Nitzschia acicularis W. Sm. and Tetraselmis suecica Butch.) Growth kinetics and flourescence
Fengbo Li et al.
Chemical communications (Cambridge, England), (17)(17), 2238-2240 (2005-04-28)
Barium sulfate nanotubes perform excellently in supporting sulfates (Pt(SO4)2, HgSO4, Ce(SO4)2 and Pb(SO4)2) for low temperature catalytic conversion of methane to methanol under strongly acidic conditions in a conventional gas-phase reactor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service