Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V800253

Sigma-Aldrich

Mercury(II) chloride

LR, ≥99%

Synonym(s):

Mercuric chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgCl2
CAS Number:
Molecular Weight:
271.50
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99%
grade:
LR
form:
crystals
Pricing and availability is not currently available.

grade

LR

vapor pressure

1.3 mmHg ( 236 °C)

product line

Vetec

assay

≥99%

form

crystals

mp

277 °C (lit.)

SMILES string

Cl[Hg]Cl

InChI

1S/2ClH.Hg/h2*1H;/q;;+2/p-2

InChI key

LWJROJCJINYWOX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philipp Lehner et al.
PloS one, 10(6), e0128125-e0128125 (2015-06-02)
Most commercially available optical oxygen sensors target the measuring range of 300 to 2 μmol L-1. However these are not suitable for investigating the nanomolar range which is relevant for many important environmental situations. We therefore developed a miniaturized phase
Kshitiz Tyagi et al.
Nucleic acids research, 43(9), 4701-4712 (2015-04-15)
Maintenance of protein quality control has implications in various processes such as neurodegeneration and ageing. To investigate how environmental insults affect this process, we analysed the proteome of yeast continuously exposed to mild heat stress. In agreement with previous transcriptomics
Liakot A Khan et al.
Nature cell biology, 15(2), 143-156 (2013-01-22)
Many unicellular tubes such as capillaries form lumens intracellularly, a process that is not well understood. Here we show that the cortical membrane organizer ERM-1 is required to expand the intracellular apical/lumenal membrane and its actin undercoat during single-cell Caenorhabditis
Andrés Cruz Hernández et al.
Metallomics : integrated biometal science, 5(4), 398-403 (2013-03-27)
RNA processing is an essential pathway in the regulation of genetic expression in the cell. In this work, Bacillus subtilis was used to understand the effects of mercury on the mechanism of tRNA metabolism. The CVAAS (cold vapor atomic absorption
Michael Gülden et al.
Toxicology, 332, 41-51 (2013-12-03)
In vitro generated data on toxic potencies are generally based on nominal concentrations. However, cellular and extracellular binding and elimination processes may reduce the available free fraction of a compound. Then, nominal effective concentrations do not represent appropriate measures of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service