Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V800251

Sigma-Aldrich

Mercury(II) acetate

AR, ≥99%

Synonym(s):

Mercuric acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Hg
CAS Number:
Molecular Weight:
318.68
Beilstein/REAXYS Number:
3563831
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
assay:
≥99%
grade:
AR
form:
powder
Pricing and availability is not currently available.

grade

AR

product line

Vetec

assay

≥99%

form

powder

mp

179-182 °C (lit.)

anion traces

chloride (Cl-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.002%

SMILES string

CC(=O)O[Hg]OC(C)=O

InChI

1S/2C2H4O2.Hg/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

BRMYZIKAHFEUFJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Gellerstedt et al.
Carbohydrate research, 294, 41-51 (1996-11-20)
It has recently been demonstrated that 4-deoxy-L-threo-hex-4-enopyranosyluronic acid ("hexeneuronic acid") is present in kraft pulps and linked to the xylan backbone. An analytical method for the quantitative determination of hexeneuronic acid groups has now been developed. The procedure involves a
Z Gregus et al.
Toxicology and applied pharmacology, 174(2), 177-187 (2001-07-12)
Interaction of methylmercury (MM), an environmental and industrial toxicant, with selenium is well known but incompletely understood. Therefore, the effects of MM (10 micromol/kg i.v.) on the disposition of exogenous selenium were compared with those of other organic mercurials (merbromine
R I Boysen et al.
The journal of peptide research : official journal of the American Peptide Society, 57(1), 19-28 (2001-02-13)
The TthL36 protein of the 50S ribosomal proteins from Thermus thermophilus has been found to contain the rare C(Xaa)2C(Xaa)12C(Xaa)4H (CCCH) sequence motif, a zinc finger binding motif, which for other zinc finger proteins is known to cleave RNA hairpins. In
N Milosevic et al.
European journal of pharmacology, 401(3), 317-328 (2000-08-11)
The role of intercellular signalling between liver cells in lead (Pb)(1)-induced liver toxicity was investigated in cocultures of freshly isolated and cultured rat hepatocytes and Kupffer cells. The Kupffer cells (seeded onto culture dish inserts), the hepatocytes or the two
Y Hojima et al.
Matrix biology : journal of the International Society for Matrix Biology, 14(2), 113-120 (1994-03-01)
Procollagen C- and N-proteinases specifically cleave the C- and N-terminal extension propeptides of type I, II and III procollagen molecules. The collagen molecules generated by the enzymes self-assemble into collagen fibrils. We previously observed the inhibition of these enzymes purified

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service