Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800239

Sigma-Aldrich

Paraffin oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352300
Pricing and availability is not currently available.

product line

Vetec

Quality Level

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

419.0 °F - closed cup

flash_point_c

215 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Schmidts et al.
International journal of pharmaceutics, 398(1-2), 107-113 (2010-08-03)
Multiple water-in-oil-in-water (W/O/W) emulsions are of major interest as potential skin delivery systems for water-soluble drugs like oligonucleotides due to their distinct encapsulation properties. However, multiple emulsions are highly sensitive in terms of variations of the individual components. The presence
Characteristic adverse skin reactions to antiseptic bath oils.
Mignon Moyle et al.
The Medical journal of Australia, 186(12), 652-653 (2007-06-20)
Ozlem Yigit et al.
The Journal of emergency medicine, 42(4), 417-419 (2010-12-24)
Fire-eater's pneumonia is a chemical pneumonitis that can develop after accidental aspiration of liquid hydrocarbon-based fuel during a flame-blowing or a fire-eating performance. Typical findings of the patient are similar with any infectious pneumonia: chest pain, shortness of breath, cough
Nicole Concin et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46(2), 544-552 (2007-10-10)
Paraffins of mineral oil origin (mineral paraffins) were analyzed in tissue fat collected from 144 volunteers with Caesarean sections as well as in milk fat from days 4 and 20 after birth of the same women living in Austria. In
Joo-Hark Yi et al.
Clinical journal of the American Society of Nephrology : CJASN, 7(8), 1211-1216 (2012-06-16)
For accurate measurement of pH, urine collection under oil to limit the escape of CO(2) on air exposure is recommended. This study aims to test the hypothesis that urine collection under oil is not necessary in acidic urine in which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service