Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800230

Sigma-Aldrich

DL-Lactic acid

AR, ≥88%

Synonym(s):

2-Hydroxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)COOH
CAS Number:
Molecular Weight:
90.08
Beilstein/REAXYS Number:
1209341
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

AR

product line

Vetec

assay

≥88%

form

viscous liquid

refractive index

n20/D 1.425 (lit.)

pH

2

bp

122 °C/15 mmHg (lit.)

density

1.209 g/mL at 25 °C (lit.)

SMILES string

CC(O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)

InChI key

JVTAAEKCZFNVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Eileen F O' Shea et al.
Current opinion in biotechnology, 24(2), 130-134 (2013-01-23)
Lactic acid bacteria (LAB) produce a wide variety of antimicrobial peptides (bacteriocins) which contribute to the safety and preservation of fermented foods. This review discusses strategies that have been or could be employed to further enhance the commercial application of
Paula Gaspar et al.
Biotechnology advances, 31(6), 764-788 (2013-04-10)
The lactic acid bacteria (LAB) are a functionally related group of low-GC Gram-positive bacteria known essentially for their roles in bioprocessing of foods and animal feeds. Due to extensive industrial use and enormous economical value, LAB have been intensively studied
Annabelle Olsson et al.
Journal of wildlife diseases, 49(3), 560-567 (2013-06-20)
Using a prospective, randomized study design we demonstrate that midazolam sedation minimizes acidosis compared with physical restraint in captive juvenile estuarine crocodiles during handling or noninvasive procedures at preferred body temperature. A dose of midazolam (5.0 mg/kg) was administered intramuscularly
Jan Martinussen et al.
Current opinion in biotechnology, 24(2), 124-129 (2012-12-26)
The ability of lactic acid bacteria to produce lactic acid from various sugars plays an important role in food fermentations. Lactic acid is derived from pyruvate, the end product of glycolysis and thus a fast lactic acid production rate requires
James Steele et al.
Current opinion in biotechnology, 24(2), 135-141 (2013-01-03)
It has been known since the 1960s that lactic acid bacteria are essential for the development of cheese flavor. In the ensuing 50 years significant research has been directed at understanding the microbiology, genetics and biochemistry of this process. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service