Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800221

Sigma-Aldrich

4-Methyl-2-pentanone

AR, ≥99%

Synonym(s):

Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCH3
CAS Number:
Molecular Weight:
100.16
Beilstein/REAXYS Number:
605399
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
AR
assay:
≥99%
bp:
117-118 °C
Pricing and availability is not currently available.

grade

AR

vapor density

3.5 (vs air)

product line

Vetec

assay

≥99%

form

liquid

autoignition temp.

840 °F

expl. lim.

1.2-8 %, 93 °F

dilution

(for analytical testing)

refractive index

n20/D 1.395 (lit.)

bp

117-118 °C

mp

−80 °C (lit.)

density

0.801 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(C)=O

InChI

1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3

InChI key

NTIZESTWPVYFNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

57.2 °F - closed cup

flash_point_c

14 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H J Shim et al.
Journal of chromatography. B, Biomedical applications, 656(2), 407-414 (1994-06-17)
A high-performance liquid chromatographic method was developed for the simultaneous determination of a new anthracycline, DA-125 (I), and its metabolites (M1, M2, M3, and M4) in rat plasma and urine using fluorescein as an internal standard. Compound I, a prodrug
Ginette Truchon et al.
Journal of occupational and environmental hygiene, 3(3), 137-143 (2006-02-09)
Compartmental and physiologically based toxicokinetic modeling coupled with Monte Carlo simulation were used to quantify the impact of biological variability (physiological, biochemical, and anatomic parameters) on the values of a series of bio-indicators of metal and organic industrial chemical exposures.
David Quesnel et al.
Water research, 39(4), 677-687 (2005-02-15)
An industrial wastewater that was pretreated by an aerobic thermophilic bacterial consortium (THE) was subjected to additional treatability studies by granular activated carbon (GAC) and a conventional activated sludge (CAS). The removal of dissolved organic carbon (DOC) in both systems
Mao Ting et al.
The Science of the total environment, 390(1), 97-108 (2007-11-03)
Volatile organic compounds (VOCs) in the air of Beijing City were measured at the heights of 8, 32, 140 and 280 m on the Beijing 325 m meteorological tower in autumn 2005. Concentrations of fifty-five compounds were determined by quantitative
Ting-Ke Tseng et al.
Journal of hazardous materials, 122(1-2), 155-160 (2005-05-26)
Catalytic oxidation is one of the cost-effective technologies to solve the troublesome volatile organic compounds. This study treated methyl-isobutyl-ketone (MIBK) by a commercial catalyst, Pt/gamma-Al(2)O(3), in a fixed-bed reactor. The effects of operating factors, such as operating temperature, MIBK concentration

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service