Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800213

Sigma-Aldrich

Hydroquinone

LR, ≥99%

Synonym(s):

1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein/REAXYS Number:
605970
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

LR

vapor density

3.81 (vs air)

vapor pressure

1 mmHg ( 132 °C)

product line

Vetec

assay

≥99%

autoignition temp.

930 °F

bp

285 °C (lit.)

mp

172-175 °C (lit.)

SMILES string

Oc1ccc(O)cc1

InChI

1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H

InChI key

QIGBRXMKCJKVMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1B

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

329.0 °F - closed cup

flash_point_c

165 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaqi Hu et al.
Biosensors & bioelectronics, 47, 45-49 (2013-04-03)
A novel light "on-off" photoelectrochemical sensing for the sensitive determination of hydroquinone was developed based on the porphyrin-functionalized Au nanoparticles on graphene (porphyrin/AuNPs/graphene). Gold nanoparticles (AuNPs) were firstly modified onto graphene sheets using NaBH4 as reductant and the porphyrin/AuNPs/graphene nanocomposites
P Carbonnelle et al.
Toxicology and applied pharmacology, 132(2), 220-226 (1995-06-01)
Decreased interleukin-1 (IL-1) production by mononuclear phagocytes has been shown to contribute to benzene myelotoxicity in animals. The study presented here was designed to examine the relevance of this mechanism in humans. Fresh human blood monocytes were exposed to 0.001-10
Douglas McGregor
Critical reviews in toxicology, 37(10), 887-914 (2007-11-21)
The toxicology of hydroquinone has been reviewed on a number of previous occasions. This review targets its potential for carcinogenicity and possible modes of carcinogenic action. The evaluation made by IARC (1999) of its carcinogenic risk to humans was that
A P DeCaprio
Critical reviews in toxicology, 29(3), 283-330 (1999-06-24)
Hydroquinone (HQ) is a high-volume commodity chemical used as a reducing agent, antioxidant, polymerization inhibitor, and chemical intermediate. It is also used in over-the-counter (OTC) drugs as an ingredient in skin lighteners and is a natural ingredient in many plant-derived
Xia Cao et al.
Analytica chimica acta, 752, 101-105 (2012-10-30)
Ultrathin crystalline CdSe nanosheets have been synthesized through a facile solution processes. Their application to simultaneous electrochemical determination of catechol and hydroquinone is demonstrated. The few-layer single crystalline CdSe modified electrode exhibits strong electrocatalytic activity toward the oxidation of a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service