Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800173

Sigma-Aldrich

Ethylenediaminetetraacetic acid solution

0.02 N

Synonym(s):

EDTA solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O8
CAS Number:
Molecular Weight:
292.24
Beilstein/REAXYS Number:
1716295
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Pricing and availability is not currently available.

product line

Vetec

form

liquid

concentration

0.02 N

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)

InChI key

KCXVZYZYPLLWCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucienne Tritten et al.
PLoS neglected tropical diseases, 8(7), e2971-e2971 (2014-07-18)
Filarial nematodes cause chronic and profoundly debilitating diseases in both humans and animals. Applications of novel technology are providing unprecedented opportunities to improve diagnosis and our understanding of the molecular basis for host-parasite interactions. As a first step, we investigated
Derek K Chu et al.
The Journal of experimental medicine, 211(8), 1657-1672 (2014-07-30)
Eosinophils natively inhabit the small intestine, but a functional role for them there has remained elusive. Here, we show that eosinophil-deficient mice were protected from induction of Th2-mediated peanut food allergy and anaphylaxis, and Th2 priming was restored by reconstitution
Khosrow Adeli et al.
Clinical chemistry, 61(8), 1063-1074 (2015-06-06)
Defining laboratory biomarker reference values in a healthy population and understanding the fluctuations in biomarker concentrations throughout life and between sexes are critical to clinical interpretation of laboratory test results in different disease states. The Canadian Health Measures Survey (CHMS)
Yoshihiro Yoshitake et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(2), 312-321 (2014-11-14)
The peptides derived from ideal cancer-testis antigens, including LY6K, CDCA1, and IMP3 (identified using genome-wide cDNA microarray analyses), were used in immunotherapy for head and neck squamous cell cancer (HNSCC). In this trial, we analyzed the immune response to and
Cecilia Lezcano et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 27(9), 1193-1202 (2014-01-18)
Of more than 150 000 published studies evaluating new biomarkers, fewer than 100 biomarkers have been implemented for patient care. One reason for this is lack of rigorous testing by the medical community to validate claims for biomarker clinical relevance, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service