Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800163

Sigma-Aldrich

Dimethyl sulfoxide

LR, ≥99%

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein/REAXYS Number:
506008
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
LR
assay:
≥99%
bp:
189 °C (lit.)
vapor pressure:
0.42 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

LR

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

product line

Vetec

assay

≥99%

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

dilution

(for analytical testing)

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

density

1.10 g/mL (lit.)

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongjie An et al.
Advances in colloid and interface science, 222, 9-17 (2014-08-17)
The observation by Atomic Force Microscopy of a range of nanophases on hydrophobic surfaces poses some challenging questions, not only related to the stability of these objects but also regarding their wetting properties. Spherical capped nanobubbles are observed to exhibit
A Oguro-Ando et al.
Molecular psychiatry, 20(9), 1069-1078 (2014-10-15)
Rare maternally inherited duplications at 15q11-13 are observed in ~1% of individuals with an autism spectrum disorder (ASD), making it among the most common causes of ASD. 15q11-13 comprises a complex region, and as this copy number variation encompasses many
Claudia C Faria et al.
Cancer research, 75(1), 134-146 (2014-11-14)
Medulloblastoma is the most common malignant pediatric brain tumor, with metastases present at diagnosis conferring a poor prognosis. Mechanisms of dissemination are poorly understood and metastatic lesions are genetically divergent from the matched primary tumor. Effective and less toxic therapies
Björn Zörner et al.
Brain : a journal of neurology, 137(Pt 6), 1716-1732 (2014-04-17)
Anatomical plasticity such as fibre growth and the formation of new connections in the cortex and spinal cord is one known mechanism mediating functional recovery after damage to the central nervous system. Little is known about anatomical plasticity in the
Mulmudi Hemant Kumar et al.
Advanced materials (Deerfield Beach, Fla.), 26(41), 7122-7127 (2014-09-13)
Lead free perovskite solar cells based on a CsSnI3 light absorber with a spectral response from 950 nm is demonstrated. The high photocurrents noted in the system are a consequence of SnF2 addition which reduces defect concentrations and hence the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service