Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800143

Sigma-Aldrich

D-(+)-Glucose

LR, anhydrous, ≥99.5%

Synonym(s):

Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein/REAXYS Number:
1724615
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Pricing and availability is not currently available.

grade

LR
anhydrous

product line

Vetec

assay

≥99.5%

mp

150-152 °C (lit.)

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jashanpreet Kaur et al.
PloS one, 10(3), e0122354-e0122354 (2015-03-27)
Genotyping studies of Australian Scedosporium isolates have revealed the strong prevalence of a recently described species: Scedosporium aurantiacum. In addition to occurring in the environment, this fungus is also known to colonise the respiratory tracts of cystic fibrosis (CF) patients.
Ragadeepthi Tunduguru et al.
Biochemical pharmacology, 92(2), 380-388 (2014-09-10)
Skeletal muscle accounts for ∼ 80% of postprandial glucose clearance, and skeletal muscle glucose clearance is crucial for maintaining insulin sensitivity and euglycemia. Insulin-stimulated glucose clearance/uptake entails recruitment of glucose transporter 4 (GLUT4) to the plasma membrane (PM) in a
Loes P M Duivenvoorde et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 70(3), 282-293 (2014-03-13)
Indirect calorimetry (InCa) can potentially be used to noninvasively assess metabolic and age-related flexibility. To assess the use of InCa for this purpose, we tested the sensitivity and response stability over time of three InCa-based treatments in old versus adult
Aïcha Gharbi et al.
AMB Express, 5, 9-9 (2015-04-09)
Carvacrol, an aromatic terpenic compound, known to be antimicrobial was grafted onto gold surfaces via two strategies based on newly-synthesized cross-linkers involving either an ester bond which can be cleaved by microbial esterases, or a covalent ether link. Surface functionalizations
Yu-ting Alex Chiang et al.
American journal of physiology. Endocrinology and metabolism, 306(7), E707-E722 (2013-12-26)
p21-Activated protein kinases (PAKs) are centrally involved in a plethora of cellular processes and functions. Their function as effectors of small GTPases Rac1 and Cdc42 has been extensively studied during the past two decades, particularly in the realms of cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service