Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800135

Sigma-Aldrich

Copper(I) chloride

LR, ≥96%

Synonym(s):

Copper monochloride, Cuprous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl
CAS Number:
Molecular Weight:
99.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥96%
grade:
LR
form:
crystalline powder
Pricing and availability is not currently available.

grade

LR

vapor pressure

1.3 mmHg ( 546 °C)

product line

Vetec

assay

≥96%

form

crystalline powder

pH

5 (20 °C, 50 g/L)

bp

1490 °C (lit.)

mp

430 °C (lit.)

SMILES string

Cl[Cu]

InChI

1S/ClH.Cu/h1H;/q;+1/p-1

InChI key

OXBLHERUFWYNTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Shows unique character as an initiator of radical reactions such as the hydrostannation of α,β-unsaturated ketones.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Fan Zhao et al.
Journal of colloid and interface science, 448, 380-388 (2015-03-11)
Here we describe the development of versatile antifouling polyethersulfone (PES) filtration membranes modified via surface grafting of zwitterionic polymers from a reactive amphiphilic copolymer additive. Amphiphilic polyethersulfone-block-poly(2-hydroxyethyl methacrylate) (PES-b-PHEMA) was beforehand designed and used as the blending additive of PES
Yew-Foon Tan et al.
Journal of proteome research, 11(7), 3860-3879 (2012-05-12)
Plant mitochondria are highly responsive organelles that vary their metabolism in response to a wide range of chemical and environmental conditions. Quantitative proteomics studies have begun to allow the analysis of these large-scale protein changes in mitochondria. However studies of
Marie Sircoglou et al.
Journal of the American Chemical Society, 130(49), 16729-16738 (2009-06-26)
The ambiphilic triphosphine-borane ligand 1 {TPB = [o-iPr2P-(C6H4)3B} readily coordinates to all group 10 and 11 metals to afford a complete series of metal boratranes (TPB)[M] 2-8 (2: M = Ni, 3: M = Pd, 4: M = Pt, 5:
Haiyuan Zhang et al.
Academic radiology, 18(12), 1561-1568 (2011-11-08)
The aim of this study was to assess copper metabolism of human hepatocellular carcinoma (HCC) with positron emission tomographic (PET) imaging using copper (II)-64 chloride ((64)CuCl(2)) as a tracer. PET imaging of athymic mice (n = 5) bearing extrahepatic HCC
Manuela Natoli et al.
Journal of cellular physiology, 226(6), 1531-1543 (2010-10-15)
The human intestinal Caco-2 cell line has been extensively used as a model of the intestinal barrier. However, it is widely reported in literature that culture-related conditions, as well as the different Caco-2 cell lines utilized in different laboratories, often

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service