Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800116

Sigma-Aldrich

Chloroform

LR, contains 100 ppm amylene as stabilizer, ≥99%

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein/REAXYS Number:
1731042
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
LR
assay:
≥99%
bp:
60.5-61.5 °C (lit.)
vapor pressure:
160 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

LR

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

product line

Vetec

assay

≥99%

form

liquid

contains

100 ppm amylene as stabilizer

dilution

(for analytical testing)

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Accalia Fu et al.
Diabetologia, 58(7), 1513-1522 (2015-04-16)
Precise regulation of insulin secretion by the pancreatic beta cell is essential for the maintenance of glucose homeostasis. Insulin secretory activity is initiated by the stepwise breakdown of ambient glucose to increase cellular ATP via glycolysis and mitochondrial respiration. Knockout
Kuan Sun et al.
Nature communications, 6, 6013-6013 (2015-01-15)
Solution-processed organic photovoltaic cells (OPVs) hold great promise to enable roll-to-roll printing of environmentally friendly, mechanically flexible and cost-effective photovoltaic devices. Nevertheless, many high-performing systems show best power conversion efficiencies (PCEs) with a thin active layer (thickness is ~100 nm) that
Todd A Lydic et al.
Methods (San Diego, Calif.), 87, 83-95 (2015-04-25)
There is an increasing recognition of the role that cancer cell derived exosomes play in intercellular signaling upon fusion or uptake with a target cell, including immune system evasion, tumor growth and metastasis. To date, however, although exosomal membrane and
Shuiquan Tang et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 368(1616), 20120318-20120318 (2013-03-13)
Two novel reductive dehalogenases (RDases) that are highly similar to each other but catalyse distinct dechlorination reactions were identified from Dehalobacter-containing mixed cultures. These two RDases were partially purified from crude protein extracts of anaerobic dechlorinating enrichment cultures using blue
Pramod Kumar et al.
Tissue engineering. Part C, Methods, 21(7), 660-670 (2014-12-24)
Tissue engineering by self-assembly uses the cells' secretome as a regeneration template and biological factory of trophic factors. Despite the several advantages that have been witnessed in preclinical and clinical settings, the major obstacle for wide acceptance of this technology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service