Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800113

Sigma-Aldrich

Hexadecyltrimethylammonium bromide

AR, ≥99%

Synonym(s):

CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(Br)(CH3)3
CAS Number:
Molecular Weight:
364.45
Beilstein/REAXYS Number:
3598189
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Pricing and availability is not currently available.

grade

AR

product line

Vetec

assay

≥99%

mol wt

364.45 g/mol

mp

248-251 °C (lit.)

SMILES string

[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1

InChI key

LZZYPRNAOMGNLH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

target_organs

Gastrointestinal tract, Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

471.2 °F - closed cup

flash_point_c

244 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fay-Wei Li et al.
Nature communications, 6, 7852-7852 (2015-07-29)
Phytochromes are red/far-red photoreceptors that play essential roles in diverse plant morphogenetic and physiological responses to light. Despite their functional significance, phytochrome diversity and evolution across photosynthetic eukaryotes remain poorly understood. Using newly available transcriptomic and genomic data we show
Armando Maestro et al.
Langmuir : the ACS journal of surfaces and colloids, 31(23), 6289-6297 (2015-05-15)
We address the rheology of assemblies of surfactant-decorated silica nanoparticles irreversibly adsorbed at the gas/liquid interface. Positively charged surfactant molecules (such as CTAB) bind to silica nanoparticle surfaces, and the resulting particle-surfactant complexes adsorb at gas/liquid interfaces. The surfactant molecules
A T Nasr et al.
Physics in medicine and biology, 60(12), 4685-4704 (2015-05-29)
In this study, recipe optimization of Leuco Crystal Violet (LCV) micelle gels made with the surfactant Cetyl Trimethyl Ammonium Bromide (CTAB) and the chemical sensitizer 2,2,2-trichloroethanol (TCE) was aided by a two-level three-factor designed experiment. The optimized recipe contains 0.75
Moritz Tebbe et al.
Faraday discussions, 181, 243-260 (2015-05-08)
We demonstrate the large-scale organisation of anisotropic nanoparticles into linear assemblies displaying optical anisotropy on macroscopic areas. Monodisperse gold nanorods with a hydrophilic protein shell are arranged by dip-coating on wrinkled surfaces and subsequently transferred to indium tin oxide (ITO)
Carina Hillgruber et al.
The Journal of experimental medicine, 212(8), 1255-1266 (2015-07-15)
Spontaneous organ hemorrhage is the major complication in thrombocytopenia with a potential fatal outcome. However, the exact mechanisms regulating vascular integrity are still unknown. Here, we demonstrate that neutrophils recruited to inflammatory sites are the cellular culprits inducing thrombocytopenic tissue

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service