Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800104

Sigma-Aldrich

Calcium chloride

LR, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCl2
CAS Number:
Molecular Weight:
110.98
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥98%
grade:
LR
form:
lumps
Pricing and availability is not currently available.

grade

LR

vapor pressure

0.01 mmHg ( 20 °C)
0.01 mmHg ( 20 °C)

product line

Vetec

assay

≥98%

form

lumps

mp

772 °C (lit.)

cation traces

Fe: ≤0.01%
heavy metals (as Pb): ≤0.002%

SMILES string

[Cl-].[Cl-].[Ca++]

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dino Demirovic et al.
Journal of cell communication and signaling, 8(4), 333-339 (2014-09-07)
Repeated exposure to mild heat shock (HS) has been shown to induce a wide range of health promoting hormetic effects in various biological systems, including human cells undergoing aging in vitro. In order to understand how cells distinguish between mild
Kaisa Kyöstilä et al.
PLoS genetics, 11(4), e1005169-e1005169 (2015-04-16)
Inherited neurodegenerative disorders are debilitating diseases that occur across different species. We have performed clinical, pathological and genetic studies to characterize a novel canine neurodegenerative disease present in the Lagotto Romagnolo dog breed. Affected dogs suffer from progressive cerebellar ataxia
Matthias Eibauer et al.
Nature communications, 6, 7532-7532 (2015-06-27)
Nuclear pore complexes (NPCs) perforate the nuclear envelope and allow the exchange of macromolecules between the nucleus and the cytoplasm. To acquire a deeper understanding of this transport mechanism, we analyse the structure of the NPC scaffold and permeability barrier
Takashi Yamada et al.
Skeletal muscle, 5, 20-20 (2015-07-15)
In addition to the primary symptoms arising from inflamed joints, muscle weakness is prominent and frequent in patients with rheumatoid arthritis (RA). Here, we investigated the mechanisms of arthritis-induced muscle dysfunction in rats with adjuvant-induced arthritis (AIA). AIA was induced
Konstantinos Malliaras et al.
EMBO molecular medicine, 6(6), 760-777 (2014-05-07)
Controversy surrounds the identity, origin, and physiologic role of endogenous cardiomyocyte progenitors in adult mammals. Using an inducible genetic labeling approach to identify small non-myocyte cells expressing cardiac markers, we find that activated endogenous cardioblasts are rarely evident in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service