Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800086

Sigma-Aldrich

Bromine

LR, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Br2
CAS Number:
Molecular Weight:
159.81
EC Number:
MDL number:
UNSPSC Code:
12141915
PubChem Substance ID:
assay:
≥99%
grade:
LR
form:
liquid
Pricing and availability is not currently available.

grade

LR

vapor density

7.14 (vs air)

vapor pressure

175 mmHg ( 20 °C)
671 mmHg ( 55 °C)

product line

Vetec

assay

≥99%

form

liquid

resistivity

7.8E18 μΩ-cm, 20°C

bp

58.8 °C (lit.)

mp

−7.2 °C (lit.)

density

3.119 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.2%

SMILES string

BrBr

InChI

1S/Br2/c1-2

InChI key

GDTBXPJZTBHREO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitalij V Levin et al.
Organic letters, 15(4), 917-919 (2013-02-02)
Reactions of difluorocarbene with benzyl and alkylzinc halides leading to fluorinated organozinc species have been described. The generated α-difluorinated organozinc reagents are reasonably stable in solution and can be quenched with external electrophiles (iodine, bromine, proton), affording compounds containing the
Kimberly T Barrett et al.
Journal of the American Chemical Society, 135(8), 2963-2966 (2013-02-16)
We report the enantioselective synthesis of atropisomeric benzamides employing catalytic electrophilic aromatic substitution reactions involving bromination. The catalyst is a simple tetrapeptide bearing a tertiary amine that may function as a Brønsted base. A series of tri- and dibrominations were
Nickel-butadiene catalytic system for the cross-coupling of bromoalkanoic acids with alkyl Grignard reagents: a practical and versatile method for preparing fatty acids.
Takanori Iwasaki et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(9), 2956-2960 (2013-02-02)
Bromine-77 and iodine-123 radiopharmaceuticals.
G Stöcklin
The International journal of applied radiation and isotopes, 28(1-2), 131-147 (1977-01-01)
John D Sivey et al.
Environmental science & technology, 47(3), 1330-1338 (2013-01-18)
HOBr, formed via oxidation of bromide by free available chlorine (FAC), is frequently assumed to be the sole species responsible for generating brominated disinfection byproducts (DBPs). Our studies reveal that BrCl, Br(2), BrOCl, and Br(2)O can also serve as brominating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service