Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800077

Sigma-Aldrich

Benzoyl chloride

AR, ≥99%

Synonym(s):

Benzoic acid chloride, alpha-chlorobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCl
CAS Number:
Molecular Weight:
140.57
Beilstein/REAXYS Number:
471389
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

AR

Quality Level

vapor density

4.88 (vs air)

vapor pressure

1 mmHg ( 32 °C)

product line

Vetec

assay

≥99%

autoignition temp.

1056 °F

expl. lim.

4.9 %

refractive index

n20/D 1.553 (lit.)

bp

198 °C (lit.)

mp

−1 °C (lit.)

density

1.211 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)c1ccccc1

InChI

1S/C7H5ClO/c8-7(9)6-4-2-1-3-5-6/h1-5H

InChI key

PASDCCFISLVPSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

161.6 °F - closed cup

flash_point_c

72 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ozgül Ozdestan et al.
Talanta, 78(4-5), 1321-1326 (2009-04-14)
A new benzoylation procedure was developed which was more reliable than the most common benzoylation procedures in terms of reaction time, peak resolution, detector response and repeatability. Methylamine, putrescine, cadaverine, tryptamine, beta-phenylethylamine, spermidine, spermine, histamine, tyramine and agmatine were studied.
Cong Li et al.
Journal of the American Society for Mass Spectrometry, 24(4), 493-501 (2013-03-21)
The benzoxyl radical is a key intermediate in the combustion of toluene and other aromatic hydrocarbons, yet relatively little experimental work has been performed on this species. Here, a combination of electrospray ionization (ESI), multistage mass spectrometry experiments, and density
Russell J Cox et al.
Chemical communications (Cambridge, England), (8)(8), 1037-1039 (2005-02-19)
Conditions are reported for the facile, high-yielding coupling of acyl chlorides with terminal alkynes in a reaction involving palladium and copper iodide; the reaction is tolerant of a wide variety of acyl chlorides and terminal alkynes and provides a convenient
Ran Liu et al.
Journal of mass spectrometry : JMS, 47(10), 1341-1346 (2012-09-29)
A high-performance liquid chromatography coupled with Q-time of flight mass spectrometry (HPLC/Q-TOF MS) method was developed and validated for the determination of 1, 3-diaminopropane, putrescine, cadaverine, spermidine and spermine in human plasma. The plasma samples were first pretreated by 10%
Vutukuri Prakash Reddy et al.
Organic & biomolecular chemistry, 13(24), 6803-6813 (2015-05-27)
The direct sulfenylation and sulfonylation of (sp(2))C-H bonds of benzamide derivatives were achieved using a Ni catalyst with the aid of an 8-aminoquinoline moiety as a bidentate directing group. These protocols represent a convenient route for the formation of valuable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service