Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800066

Sigma-Aldrich

Barium hydroxide

LR, ≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(OH)2
CAS Number:
Molecular Weight:
171.34
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥97%
grade:
LR
form:
crystalline powder
Pricing and availability is not currently available.

grade

LR

product line

Vetec

assay

≥97%

form

crystalline powder

pH

12.5 (20 °C, 50 g/L)

mp

>300 °C (lit.)

density

2.2 g/mL at 25 °C (lit.)

SMILES string

O[Ba]O

InChI

1S/Ba.2H2O/h;2*1H2/q+2;;/p-2

InChI key

RQPZNWPYLFFXCP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M F Byford
The Biochemical journal, 280 ( Pt 1), 261-265 (1991-11-15)
The beta-elimination of phosphoserine residues by dilute alkali is catalysed by the presence of group II metal ions. The use of 0.1 M-Ba (OH)2 catalysed the rate of beta-elimination of phosphoserine by more than two orders of magnitude compared with
T Irie et al.
Carbohydrate research, 192, 167-172 (1989-10-23)
The alkylation of cyclomalto-oligosaccharides (cyclodextrins, CDs) with dialkyl sulfate-barium hydroxide has been claimed to yield 2,6-di-O-alkyl derivatives. Re-investigation by plasma desorption-m.s. of the products of laboratory methylation of alpha CD, beta CD, or gamma CD and ethylation of beta CD
T Mahendran et al.
Journal of agricultural and food chemistry, 56(19), 9269-9276 (2008-09-12)
The structural characteristics of the gum exudate of Acacia senegal (gum arabic) have been investigated by monitoring the composition and physicochemical properties before and after treatment with proteolytic enzyme and various alkaline systems. Molecular mass ( M w) and radius
K C Li et al.
Magnetic resonance imaging, 9(2), 141-150 (1991-01-01)
In vitro proton spectroscopy with line-width measurements and MR imaging were performed on various concentrations of commercially available single contrast (SC), double contrast, oral and rectal barium sulfate suspensions, as well as potassium sulfate, barium chloride, barium hydroxide, and 97%
H Förster et al.
Der Anaesthesist, 46(12), 1071-1075 (1998-02-06)
The various components of commercial soda lime (sodium hydroxide, potassium hydroxide, calcium hydroxide, barium hydroxide) were studied in terms of their reactivity with sevoflurane at its boiling point (59 degrees C). A simple closed system, a reflux cooler, served as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service