V800046
Ammonium iron(III) sulfate dodecahydrate
LR, ≥98%
Synonym(s):
Ammonium ferric sulfate dodecahydrate
About This Item
Recommended Products
grade
LR
product line
Vetec™
assay
≥98%
form
solid
mp
39-41 °C (lit.)
cation traces
Fe: ≤0.002%
heavy metals (as Pb): ≤0.002%
SMILES string
N.O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].OS([O-])(=O)=O.[O-]S([O-])(=O)=O
InChI
1S/Fe.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3
Inchi Key
LCPUDZUWZDSKMX-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service