Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800029

Sigma-Aldrich

Ammonium hydroxide solution

LR, ~30% NH3

Synonym(s):

Ammonia aqueous, Ammonia water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein/REAXYS Number:
3587154
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

LR

product line

Vetec

form

liquid

expl. lim.

27 %

concentration

~30% NH3

density

0.9 g/mL at 25 °C (lit.)

SMILES string

[NH4+].[OH-]

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Sang Kim et al.
International journal of molecular sciences, 20(14) (2019-07-25)
A "smart" core@shell composite nanoparticle (NP) having dual-response mechanisms (i.e., temperature and light) was synthesized, and its efficacy in the loading and release of small molecules was explored. These core@shell NPs are composed of an optically active gold nanoshell (GNS)
Annalisa Nobili et al.
Molecular neurobiology, 55(10), 7921-7940 (2018-03-01)
Imbalances between excitatory and inhibitory synaptic transmission cause brain network dysfunction and are central to the pathogenesis of neurodevelopmental disorders. Parvalbumin interneurons are highly implicated in this imbalance. Here, we probed the social behavior and hippocampal function of mice carrying
Jodi A Lee et al.
PloS one, 10(6), e0130136-e0130136 (2015-06-18)
SerpinB2 (PAI-2), a member of the clade B family of serine protease inhibitors, is one of the most upregulated proteins following cellular stress. Originally described as an inhibitor of urokinase plasminogen activator, its predominant cytoplasmic localisation suggests an intracellular function.
Konstantin Chingin et al.
Analytical chemistry, 84(15), 6856-6862 (2012-07-12)
We introduce an online multiple-junction capillary isoelectric focusing fractionator (OMJ-CIEF) for separation of biological molecules in solution by pI. In OMJ-CIEF, the separation capillary is divided into seven equal sections joined with each other via tubular Nafion membrane insertions. Each
Damien A Leach et al.
Oncotarget, 6(18), 16135-16150 (2015-05-13)
Androgen receptor (AR) signaling in stromal cells is important in prostate cancer, yet the mechanisms underpinning stromal AR contribution to disease development and progression remain unclear. Using patient-matched benign and malignant prostate samples, we show a significant association between low

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service