Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800028

Sigma-Aldrich

Aluminum sulfate hexadecahydrate

LR, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2(SO4)3 · 16H2O
CAS Number:
Molecular Weight:
630.40
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥98%
grade:
LR
form:
crystalline powder
Pricing and availability is not currently available.

grade

LR

product line

Vetec

assay

≥98%

form

crystalline powder

anion traces

chloride (Cl-): ≤0.001%

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Al.3H2O4S.16H2O/c;;3*1-5(2,3)4;;;;;;;;;;;;;;;;/h;;3*(H2,1,2,3,4);16*1H2/q2*+3;;;;;;;;;;;;;;;;;;;/p-6

InChI key

TYYRFZAVEXQXSN-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nagasathiya Krishnan et al.
European journal of mass spectrometry (Chichester, England), 21(3), 481-486 (2015-08-27)
The coordinating nature of the hydroxy carboxylic acids, such as tartaric and citric acids, has been utilized for the in-situ formation of anions representing the trivalent elements boron and aluminum and two dianions of the hydroxy acid selected under negative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service