Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800010

Sigma-Aldrich

1-Octanesulfonic acid sodium salt

suitable for HPLC, ≥99%

Synonym(s):

Octane-1-sulfonic acid sodium salt, Sodium 1-octanesulfonate, Sodium octane-1-sulfonate, Sodium octanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H17O3SNa
CAS Number:
Molecular Weight:
216.27
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99%
Pricing and availability is not currently available.

product line

Vetec

assay

≥99%

technique(s)

HPLC: suitable

SMILES string

[Na+].CCCCCCCCS([O-])(=O)=O

InChI

1S/C8H18O3S.Na/c1-2-3-4-5-6-7-8-12(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1

InChI key

HRQDCDQDOPSGBR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Ion-pairing reagent for HPLC analysis of peptides and proteins.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I T James et al.
Journal of chromatography, 525(1), 43-57 (1990-01-26)
Following a detailed study, a rapid and sensitive assay for the naturally fluorescent collagen cross-links pyridinoline and deoxypyridinoline has been developed using ion-pair reversed-phase high-performance liquid chromatography in the presence of 1-octanesulphonic acid (OSA). Pyridinoline and deoxypyridinoline were separated on
P G Jaison et al.
Journal of chromatography. A, 1216(9), 1383-1389 (2009-01-27)
Studies were carried out for the separation of uranium (U) and thorium (Th) on reversed-phase (RP) C18 columns using mandelic acid as an eluent. Retention of thorium-mandelate on the unmodified stationary phase was found to be greater than that of
Xin Su et al.
Chemical communications (Cambridge, England), 49(26), 2655-2657 (2013-02-26)
We report two CO2-responsive aqueous solutions having switchable viscosity. The solution of sodium octadecyl sulfate and 2-(dimethylamino)ethanol forms viscoelastic worm-like micelles in the presence of CO2 but not in its absence at 60 °C. Another solution of sodium stearate and
Tejwant Singh et al.
Physical chemistry chemical physics : PCCP, 12(37), 11728-11735 (2010-08-10)
We have observed dual transitions in various physical properties while investigating the surfactant-like behavior of the ionic liquid (IL) 1-butyl-3-methylimidazolium octylsulfate [C(4)mim][C(8)SO(4)] in aqueous media. Interestingly, in conductivity measurements, it was found that the second transition, which is comparatively weak
J L Razak et al.
Pharmaceutical research, 18(1), 104-111 (2001-05-05)
The aim of this study was to develop a method based upon electrokinetic chromatography (EKC) using oppositely charged surfactant vesicles as a buffer modifier to estimate hydrophobicity (log P) for a range of neutral and charged compounds. Vesicles were formed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service