Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800007

Sigma-Aldrich

Sodium 1-heptanesulfonate monohydrate

suitable for HPLC, ≥99%

Synonym(s):

1-Heptanesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH2SO3Na · H2O
CAS Number:
Molecular Weight:
220.26
Beilstein/REAXYS Number:
3731762
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99%
Pricing and availability is not currently available.

product line

Vetec

assay

≥99%

technique(s)

HPLC: suitable

functional group

sulfonic acid

SMILES string

O.[Na+].CCCCCCCS([O-])(=O)=O

InChI

1S/C7H16O3S.Na.H2O/c1-2-3-4-5-6-7-11(8,9)10;;/h2-7H2,1H3,(H,8,9,10);;1H2/q;+1;/p-1

InChI key

XWZCREJRXRKIRQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marisa Freitas et al.
Phytochemistry, 110, 91-103 (2015-01-03)
The intensification of agricultural productivity is an important challenge worldwide. However, environmental stressors can provide challenges to this intensification. The progressive occurrence of the cyanotoxins cylindrospermopsin (CYN) and microcystin-LR (MC-LR) as a potential consequence of eutrophication and climate change is
Md Iftekhar Ahmad et al.
Drug development and industrial pharmacy, 41(5), 791-800 (2014-04-04)
In this study, a dimple-shaped chitosan carrier was developed for delivering the antituberculosis drug ethambutol dihydrochloride (EDH) from a dry powder inhaler (DPI) to the lungs. Nanosized drug particles were prepared using nanospray drying. The microsized carrier was developed from
N M Kovalchuk et al.
Journal of colloid and interface science, 459, 250-256 (2015-08-25)
Mixed solutions of cationic and anionic surfactants show considerable synergism in their wetting behaviour, but their spreading is affected considerably by the phase separation processes. The valuable information about wetting properties of synergetic mixtures can be obtained by using mixtures

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service