Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800006

Sigma-Aldrich

Sodium 1-decanesulfonate

suitable for HPLC, ≥99%

Synonym(s):

1-Decanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CH2SO3Na
CAS Number:
Molecular Weight:
244.33
Beilstein/REAXYS Number:
3918920
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

product line

Vetec

assay

≥99%

technique(s)

HPLC: suitable

mp

>300 °C (lit.)

SMILES string

[Na+].CCCCCCCCCCS([O-])(=O)=O

InChI

1S/C10H22O3S.Na/c1-2-3-4-5-6-7-8-9-10-14(11,12)13;/h2-10H2,1H3,(H,11,12,13);/q;+1/p-1

InChI key

AIMUHNZKNFEZSN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Ion-associating reagent for HPLC, including analyses of peptides and proteins.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luigi Battaglia et al.
Journal of pharmaceutical sciences, 103(7), 2157-2165 (2014-05-16)
The major obstacle to glioblastoma pharmacological therapy is the overcoming of the blood-brain barrier (BBB). In literature, several strategies have been proposed to overcome the BBB: in this experimental work, solid lipid nanoparticles (SLN), prepared according to fatty acid coacervation
N M Kovalchuk et al.
Journal of colloid and interface science, 459, 250-256 (2015-08-25)
Mixed solutions of cationic and anionic surfactants show considerable synergism in their wetting behaviour, but their spreading is affected considerably by the phase separation processes. The valuable information about wetting properties of synergetic mixtures can be obtained by using mixtures
Hongyan Yang et al.
ACS chemical neuroscience, 6(8), 1487-1501 (2015-07-15)
In vivo microdialysis is widely used to investigate how neurotransmitter levels in the brain respond to biologically relevant challenges. Here, we combined recent improvements in the temporal resolution of online sampling and analysis for serotonin with a brief high-K(+) stimulus
Franziska Mohr et al.
PloS one, 10(10), e0141136-e0141136 (2015-10-28)
The muscarinic M2 receptor (M2R) acts as a negative feedback regulator in central cholinergic systems. Activation of the M2 receptor limits acetylcholine (ACh) release, especially when ACh levels are increased because acetylcholinesterase (AChE) activity is acutely inhibited. Chronically high ACh
Zita Pöstényi et al.
Journal of pharmaceutical and biomedical analysis, 102, 529-534 (2014-12-03)
Validated HPLC analysis was developed in order to monitor the level of 4-fluorodeprenyl in rats. Male Wistar rats were intraperitoneally treated with 30 mg/kg of (-)-4-fluorodeprenyl. The rats were sacrificed after 5, 15, 30 and 60 min of treatment, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service