Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800001

Sigma-Aldrich

1,2-Dichlorobenzene

LR, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
Beilstein/REAXYS Number:
606078
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
grade:
LR
assay:
≥98%
bp:
178-180 °C (lit.)
vapor pressure:
1.2 mmHg ( 20 °C)
1.6 mmHg ( 35 °C)
Pricing and availability is not currently available.

grade

LR

vapor density

5.1 (vs air)

vapor pressure

1.2 mmHg ( 20 °C)
1.6 mmHg ( 35 °C)

product line

Vetec

assay

≥98%

form

liquid

autoignition temp.

1198 °F

expl. lim.

9.2 %

dilution

(for analytical testing)

refractive index

n20/D 1.551 (lit.)

bp

178-180 °C (lit.)

mp

−18-−17 °C (lit.)

density

1.306 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1Cl

InChI

1S/C6H4Cl2/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

RFFLAFLAYFXFSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

150.8 °F - closed cup

flash_point_c

66.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sungmin Chin et al.
Chemosphere, 75(9), 1206-1209 (2009-03-10)
This study examined the catalytic oxidation of 1,2-dichlorobenzene on V(2)O(5)/TiO(2) nanoparticles. The V(2)O(5)/TiO(2) nanoparticles were synthesized by the thermal decomposition of vanadium oxytripropoxide and titanium tetraisopropoxide. The effects of the synthesis conditions, such as the synthesis temperature and precursor heating
Bimalendu Adhikari et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(41), 11488-11496 (2011-09-29)
The N-terminally pyrene-conjugated oligopeptide, Py-Phe-Phe-Ala-OMe, (Py=pyrene 1-butyryl acyl) forms transparent, stable, supramolecular fluorescent organogels in various organic solvents. One of these organogels was thoroughly studied using various techniques including transmission electron microscopy (TEM), field-emission scanning electron microscopy (FESEM), atomic force
Eric Vejerano et al.
Environmental science & technology, 45(2), 589-594 (2010-12-09)
Previous studies have shown environmentally persistent free radicals (EPFRs) form when chlorine- and hydroxy-substituted benzenes chemisorb on Cu(II)O-containing surfaces under postcombustion conditions. This paper reports the formation of EPFRs on silica particles containing 5% Fe(III)(2)O(3). The EPFRs are formed by
Eranda Maligaspe et al.
The journal of physical chemistry. A, 114(1), 268-277 (2009-11-26)
Singlet-singlet energy transfer in self-assembled via axial coordination of imidazole-appended (at different positions of one of the meso-phenyl entities) free-base tetraphenylporphyrin, H(2)PIm, to either zinc phthalocyanine, ZnPc, or zinc naphthalocyanine, ZnNc, dyads is investigated in noncoordinating solvents, o-dichlorobenzene and toluene
Ivan Guryanov et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(46), 12837-12845 (2009-10-23)
The effect of microwave (MW) irradiation and ionic liquids (IL) on the cycloaddition of azomethine ylides to [60]fullerene has been investigated by screening the reaction protocol with regard to the IL medium composition, the applied MW power, and the simultaneous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service