Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V001702

Supelco

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23151817
Pricing and availability is not currently available.

grade

high-purity grade

Quality Level

form

powder

technique(s)

LPLC: suitable

surface area

500 m2/g , approx.

matrix

Silica

matrix active group

silica

particle size

25 μm

pore size

60 Å mean pore size

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hola, este cas está dentro de la lista de polimeros aceptados en el decreto 847/2011?

    1 answer
    1. This product is intended for research use only and not animal or human consumption. For more information on our product regulations, please navigate to our Quality & Regulatory page to view our regulatory focus areas: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/regulatory-focus-areas

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service